Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 198057431-198058013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18DP
Entrez Gene ID
348840
Gene Name
ankyrin repeat domain 18D, pseudogene
Gene Aliases
-
UniGene
Hs.512227
Gene Chromosome Location
Chr.3: 198057531-198080671 on Build GRCh38
Transcripts
NR_003291.1

SNP Detail

SNP ID Chromosome Location
rs1050035609 Chr.3: 198057620-198057620 on Build GRCh38
rs573021588 Chr.3: 198057921-198057921 on Build GRCh38
rs982041377 Chr.3: 198057508-198057508 on Build GRCh38
rs539878807 Chr.3: 198057937-198057937 on Build GRCh38
rs981563134 Chr.3: 198057788-198057788 on Build GRCh38
rs993293359 Chr.3: 198057715-198057715 on Build GRCh38
rs143839439 Chr.3: 198057648-198057648 on Build GRCh38
rs79900121 Chr.3: 198057483-198057483 on Build GRCh38
rs142637308 Chr.3: 198057936-198057936 on Build GRCh38
rs868592135 Chr.3: 198057772-198057772 on Build GRCh38
rs924651233 Chr.3: 198057797-198057797 on Build GRCh38
rs556972315 Chr.3: 198057652-198057652 on Build GRCh38
rs990268497 Chr.3: 198057853-198057853 on Build GRCh38
rs549915847 Chr.3: 198057597-198057597 on Build GRCh38
rs914822901 Chr.3: 198057574-198057574 on Build GRCh38
rs571269462 Chr.3: 198057605-198057605 on Build GRCh38
rs186696558 Chr.3: 198057932-198057932 on Build GRCh38
rs148454286 Chr.3: 198057880-198057880 on Build GRCh38
rs553668504 Chr.3: 198057653-198057653 on Build GRCh38
rs557313477 Chr.3: 198057742-198057742 on Build GRCh38
rs1040129832 Chr.3: 198057941-198057941 on Build GRCh38
rs934790818 Chr.3: 198057560-198057560 on Build GRCh38
rs956270876 Chr.3: 198057839-198057839 on Build GRCh38
rs1022955130 Chr.3: 198057768-198057768 on Build GRCh38
rs181232942 Chr.3: 198057728-198057728 on Build GRCh38
rs73891822 Chr.3: 198057705-198057705 on Build GRCh38
rs542313005 Chr.3: 198057907-198057907 on Build GRCh38
rs924822593 Chr.3: 198057547-198057547 on Build GRCh38
rs1027614288 Chr.3: 198057724-198057724 on Build GRCh38
rs916033132 Chr.3: 198057473-198057473 on Build GRCh38
rs530946791 Chr.3: 198057467-198057467 on Build GRCh38
rs946417970 Chr.3: 198057603-198057603 on Build GRCh38
rs751347818 Chr.3: 198057734-198057735 on Build GRCh38
rs888884993 Chr.3: 198057633-198057633 on Build GRCh38
rs370568563 Chr.3: 198057501-198057501 on Build GRCh38
rs897568652 Chr.3: 198057971-198057971 on Build GRCh38
rs567616719 Chr.3: 198057569-198057569 on Build GRCh38

View Full Product Details