Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3942156-3942733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM86EP
Entrez Gene ID
348926
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
-
UniGene
Hs.591720
Gene Chromosome Location
Chr.4: 3941760-3955428 on Build GRCh38
Transcripts
NR_130740.1 NR_130741.1 NR_130742.1

SNP Detail

SNP ID Chromosome Location
rs939620781 Chr.4: 3942519-3942519 on Build GRCh38
rs7680741 Chr.4: 3942293-3942293 on Build GRCh38
rs915051139 Chr.4: 3942184-3942184 on Build GRCh38
rs532651229 Chr.4: 3942216-3942216 on Build GRCh38
rs558875265 Chr.4: 3942239-3942239 on Build GRCh38
rs972354458 Chr.4: 3942438-3942438 on Build GRCh38
rs199863480 Chr.4: 3942527-3942527 on Build GRCh38
rs543958298 Chr.4: 3942594-3942594 on Build GRCh38
rs374884550 Chr.4: 3942308-3942308 on Build GRCh38
rs199921853 Chr.4: 3942433-3942433 on Build GRCh38
rs953334368 Chr.4: 3942185-3942185 on Build GRCh38
rs7695071 Chr.4: 3942257-3942257 on Build GRCh38
rs140290081 Chr.4: 3942322-3942322 on Build GRCh38
rs544962468 Chr.4: 3942543-3942543 on Build GRCh38
rs948176516 Chr.4: 3942336-3942336 on Build GRCh38
rs372072043 Chr.4: 3942500-3942500 on Build GRCh38
rs542213004 Chr.4: 3942695-3942695 on Build GRCh38
rs73793815 Chr.4: 3942658-3942658 on Build GRCh38
rs377333504 Chr.4: 3942244-3942244 on Build GRCh38
rs1026624159 Chr.4: 3942606-3942606 on Build GRCh38
rs918710834 Chr.4: 3942235-3942235 on Build GRCh38
rs192044204 Chr.4: 3942245-3942245 on Build GRCh38
rs34752642 Chr.4: 3942201-3942201 on Build GRCh38
rs1041428131 Chr.4: 3942229-3942229 on Build GRCh38
rs1007263830 Chr.4: 3942574-3942574 on Build GRCh38
rs780758430 Chr.4: 3942328-3942328 on Build GRCh38
rs1011069310 Chr.4: 3942289-3942289 on Build GRCh38
rs909989481 Chr.4: 3942456-3942456 on Build GRCh38
rs565154434 Chr.4: 3942550-3942550 on Build GRCh38
rs550520488 Chr.4: 3942343-3942343 on Build GRCh38
rs149909307 Chr.4: 3942652-3942652 on Build GRCh38
rs371060373 Chr.4: 3942190-3942190 on Build GRCh38
rs112537318 Chr.4: 3942660-3942660 on Build GRCh38
rs73793813 Chr.4: 3942494-3942494 on Build GRCh38
rs183985771 Chr.4: 3942597-3942597 on Build GRCh38
rs546148273 Chr.4: 3942372-3942372 on Build GRCh38
rs147328893 Chr.4: 3942305-3942305 on Build GRCh38
rs892086607 Chr.4: 3942375-3942375 on Build GRCh38
rs200680905 Chr.4: 3942418-3942418 on Build GRCh38
rs532097992 Chr.4: 3942647-3942647 on Build GRCh38
rs545330501 Chr.4: 3942314-3942314 on Build GRCh38
rs546336910 Chr.4: 3942611-3942611 on Build GRCh38
rs369289510 Chr.4: 3942562-3942562 on Build GRCh38
rs1003789257 Chr.4: 3942362-3942362 on Build GRCh38
rs1012509133 Chr.4: 3942250-3942250 on Build GRCh38
rs911235791 Chr.4: 3942192-3942192 on Build GRCh38
rs147372971 Chr.4: 3942384-3942384 on Build GRCh38
rs879188384 Chr.4: 3942495-3942495 on Build GRCh38
rs530738729 Chr.4: 3942567-3942567 on Build GRCh38
rs186931658 Chr.4: 3942604-3942604 on Build GRCh38
rs986029186 Chr.4: 3942298-3942298 on Build GRCh38
rs555930103 Chr.4: 3942423-3942423 on Build GRCh38
rs569464738 Chr.4: 3942422-3942422 on Build GRCh38
rs148143860 Chr.4: 3942246-3942246 on Build GRCh38
rs969499966 Chr.4: 3942639-3942639 on Build GRCh38
rs538405046 Chr.4: 3942432-3942432 on Build GRCh38
rs1003233991 Chr.4: 3942258-3942258 on Build GRCh38
rs956516297 Chr.4: 3942285-3942285 on Build GRCh38
rs1031071815 Chr.4: 3942265-3942265 on Build GRCh38
rs115011982 Chr.4: 3942450-3942450 on Build GRCh38
rs34274024 Chr.4: 3942526-3942526 on Build GRCh38
rs910062782 Chr.4: 3942335-3942335 on Build GRCh38
rs1045310705 Chr.4: 3942253-3942253 on Build GRCh38
rs930044904 Chr.4: 3942241-3942241 on Build GRCh38
rs980812150 Chr.4: 3942641-3942641 on Build GRCh38
rs141975452 Chr.4: 3942292-3942292 on Build GRCh38
rs1018737656 Chr.4: 3942585-3942585 on Build GRCh38
rs946399287 Chr.4: 3942547-3942547 on Build GRCh38
rs1053088250 Chr.4: 3942522-3942522 on Build GRCh38
rs1048923333 Chr.4: 3942330-3942330 on Build GRCh38
rs1016937265 Chr.4: 3942628-3942628 on Build GRCh38
rs774796234 Chr.4: 3942443-3942443 on Build GRCh38
rs980969857 Chr.4: 3942482-3942482 on Build GRCh38
rs939492398 Chr.4: 3942650-3942650 on Build GRCh38
rs34583792 Chr.4: 3942457-3942457 on Build GRCh38
rs984555394 Chr.4: 3942622-3942622 on Build GRCh38
rs538163408 Chr.4: 3942669-3942669 on Build GRCh38
rs529744151 Chr.4: 3942370-3942370 on Build GRCh38
rs1040069872 Chr.4: 3942692-3942692 on Build GRCh38
rs943132017 Chr.4: 3942680-3942680 on Build GRCh38
rs1025650139 Chr.4: 3942446-3942446 on Build GRCh38
rs1018727978 Chr.4: 3942426-3942426 on Build GRCh38
rs144253064 Chr.4: 3942377-3942377 on Build GRCh38
rs953113775 Chr.4: 3942392-3942392 on Build GRCh38
rs7676066 Chr.4: 3942299-3942299 on Build GRCh38
rs564912512 Chr.4: 3942317-3942317 on Build GRCh38
rs201328482 Chr.4: 3942419-3942419 on Build GRCh38
rs567173242 Chr.4: 3942355-3942355 on Build GRCh38
rs566115100 Chr.4: 3942643-3942643 on Build GRCh38

View Full Product Details