Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165279416-165279965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK3P
Entrez Gene ID
2713
Gene Name
glycerol kinase 3 pseudogene
Gene Aliases
GKP3, GKTB
UniGene
Hs.654557
Gene Chromosome Location
Chr.4: 165277792-165280023 on Build GRCh38
Transcripts
NR_026575.1 NM_001161521.1 NM_001161522.1 NM_007246.3 XM_011531572.2 XM_011531575.2 XM_011531576.2 XM_017007674.1 XM_017007675.1 XM_017007676.1 XM_017007677.1

Gene Symbol
KLHL2
Entrez Gene ID
11275
Gene Name
kelch like family member 2
Gene Aliases
ABP-KELCH, MAV, MAYVEN
UniGene
Hs.388668
Gene Chromosome Location
Chr.4: 165207618-165323156 on Build GRCh38
Transcripts
NR_026575.1 NM_001161521.1 NM_001161522.1 NM_007246.3 XM_011531572.2 XM_011531575.2 XM_011531576.2 XM_017007674.1 XM_017007675.1 XM_017007676.1 XM_017007677.1

SNP Detail

SNP ID Chromosome Location
rs745656719 Chr.4: 165279551-165279551 on Build GRCh38
rs1028084541 Chr.4: 165279737-165279737 on Build GRCh38
rs750386913 Chr.4: 165279680-165279680 on Build GRCh38
rs771698645 Chr.4: 165279447-165279447 on Build GRCh38
rs772179309 Chr.4: 165279677-165279677 on Build GRCh38
rs569202205 Chr.4: 165279723-165279723 on Build GRCh38
rs762183375 Chr.4: 165279458-165279458 on Build GRCh38
rs966529613 Chr.4: 165279540-165279540 on Build GRCh38
rs754561130 Chr.4: 165279631-165279631 on Build GRCh38
rs778447801 Chr.4: 165279663-165279663 on Build GRCh38
rs763426567 Chr.4: 165279578-165279578 on Build GRCh38
rs267600070 Chr.4: 165279598-165279598 on Build GRCh38
rs778738286 Chr.4: 165279538-165279538 on Build GRCh38
rs953408271 Chr.4: 165279462-165279462 on Build GRCh38
rs774558420 Chr.4: 165279668-165279668 on Build GRCh38
rs766208187 Chr.4: 165279689-165279689 on Build GRCh38
rs919690954 Chr.4: 165279900-165279900 on Build GRCh38
rs140860521 Chr.4: 165279806-165279806 on Build GRCh38
rs1049553486 Chr.4: 165279929-165279929 on Build GRCh38
rs995287935 Chr.4: 165279729-165279729 on Build GRCh38
rs916056247 Chr.4: 165279764-165279764 on Build GRCh38
rs765162280 Chr.4: 165279605-165279605 on Build GRCh38
rs970716295 Chr.4: 165279790-165279790 on Build GRCh38
rs547041799 Chr.4: 165279610-165279610 on Build GRCh38
rs1031744675 Chr.4: 165279757-165279757 on Build GRCh38
rs778648614 Chr.4: 165279443-165279443 on Build GRCh38
rs570919454 Chr.4: 165279876-165279876 on Build GRCh38
rs146458123 Chr.4: 165279744-165279744 on Build GRCh38
rs750301026 Chr.4: 165279609-165279609 on Build GRCh38
rs528581988 Chr.4: 165279576-165279576 on Build GRCh38
rs373150647 Chr.4: 165279802-165279802 on Build GRCh38
rs755109054 Chr.4: 165279490-165279490 on Build GRCh38
rs1018923927 Chr.4: 165279525-165279525 on Build GRCh38
rs753272494 Chr.4: 165279509-165279510 on Build GRCh38
rs769305792 Chr.4: 165279692-165279692 on Build GRCh38
rs374711520 Chr.4: 165279537-165279537 on Build GRCh38
rs990327015 Chr.4: 165279763-165279763 on Build GRCh38
rs780391976 Chr.4: 165279602-165279603 on Build GRCh38
rs773702529 Chr.4: 165279475-165279475 on Build GRCh38
rs150070685 Chr.4: 165279722-165279722 on Build GRCh38
rs775969307 Chr.4: 165279575-165279575 on Build GRCh38
rs746710681 Chr.4: 165279614-165279614 on Build GRCh38
rs1036765059 Chr.4: 165279888-165279888 on Build GRCh38
rs765670700 Chr.4: 165279460-165279460 on Build GRCh38
rs779783237 Chr.4: 165279612-165279612 on Build GRCh38
rs770791697 Chr.4: 165279440-165279440 on Build GRCh38
rs977951212 Chr.4: 165279752-165279752 on Build GRCh38
rs751775890 Chr.4: 165279488-165279488 on Build GRCh38
rs995417269 Chr.4: 165279444-165279444 on Build GRCh38
rs758121564 Chr.4: 165279554-165279554 on Build GRCh38
rs143342881 Chr.4: 165279853-165279853 on Build GRCh38
rs753854436 Chr.4: 165279494-165279494 on Build GRCh38
rs780825846 Chr.4: 165279644-165279644 on Build GRCh38
rs535800390 Chr.4: 165279695-165279695 on Build GRCh38
rs957551844 Chr.4: 165279759-165279759 on Build GRCh38
rs568475801 Chr.4: 165279694-165279694 on Build GRCh38
rs931041462 Chr.4: 165279916-165279916 on Build GRCh38
rs775045992 Chr.4: 165279450-165279450 on Build GRCh38
rs10020255 Chr.4: 165279670-165279670 on Build GRCh38
rs557912973 Chr.4: 165279746-165279746 on Build GRCh38
rs779691532 Chr.4: 165279560-165279560 on Build GRCh38
rs769373573 Chr.4: 165279662-165279662 on Build GRCh38
rs757286499 Chr.4: 165279601-165279601 on Build GRCh38
rs759645731 Chr.4: 165279590-165279590 on Build GRCh38
rs745568787 Chr.4: 165279446-165279446 on Build GRCh38
rs771462788 Chr.4: 165279667-165279667 on Build GRCh38
rs6831119 Chr.4: 165279751-165279751 on Build GRCh38
rs775713226 Chr.4: 165279678-165279678 on Build GRCh38
rs767990946 Chr.4: 165279457-165279457 on Build GRCh38
rs770424494 Chr.4: 165279577-165279577 on Build GRCh38
rs762844189 Chr.4: 165279688-165279688 on Build GRCh38
rs747621033 Chr.4: 165279650-165279650 on Build GRCh38
rs939772787 Chr.4: 165279700-165279700 on Build GRCh38
rs774666529 Chr.4: 165279585-165279585 on Build GRCh38
rs1047128434 Chr.4: 165279710-165279710 on Build GRCh38
rs752705954 Chr.4: 165279597-165279597 on Build GRCh38
rs763336064 Chr.4: 165279483-165279483 on Build GRCh38
rs767685586 Chr.4: 165279595-165279595 on Build GRCh38
rs746532605 Chr.4: 165279451-165279451 on Build GRCh38
rs747595795 Chr.4: 165279625-165279625 on Build GRCh38
rs534147254 Chr.4: 165279771-165279771 on Build GRCh38
rs887212362 Chr.4: 165279715-165279715 on Build GRCh38
rs749774141 Chr.4: 165279664-165279664 on Build GRCh38
rs757119494 Chr.4: 165279500-165279500 on Build GRCh38
rs766802877 Chr.4: 165279581-165279581 on Build GRCh38
rs759859182 Chr.4: 165279676-165279676 on Build GRCh38
rs760757688 Chr.4: 165279679-165279679 on Build GRCh38
rs746590969 Chr.4: 165279567-165279567 on Build GRCh38
rs1062548 Chr.4: 165279671-165279671 on Build GRCh38
rs767618917 Chr.4: 165279511-165279512 on Build GRCh38
rs889719773 Chr.4: 165279742-165279742 on Build GRCh38
rs748563557 Chr.4: 165279760-165279760 on Build GRCh38

View Full Product Details