Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55528764-55529333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP1
Entrez Gene ID
8611
Gene Name
phospholipid phosphatase 1
Gene Aliases
LLP1a, LPP1, PAP-2a, PAP2, PPAP2A
UniGene
Hs.696231
Gene Chromosome Location
Chr.5: 55424842-55535370 on Build GRCh38
Transcripts
NM_003711.3 NM_176895.2 NR_103485.1 XM_006714724.2 NR_001575.1

Gene Symbol
RNF138P1
Entrez Gene ID
379013
Gene Name
ring finger protein 138 pseudogene 1
Gene Aliases
-
UniGene
Hs.745164
Gene Chromosome Location
Chr.5: 55528842-55534542 on Build GRCh38
Transcripts
NM_003711.3 NM_176895.2 NR_103485.1 XM_006714724.2 NR_001575.1

SNP Detail

SNP ID Chromosome Location
rs4865953 Chr.5: 55529229-55529229 on Build GRCh38
rs527710524 Chr.5: 55529080-55529080 on Build GRCh38
rs74899495 Chr.5: 55529280-55529281 on Build GRCh38
rs756303050 Chr.5: 55529093-55529093 on Build GRCh38
rs866611935 Chr.5: 55528862-55528862 on Build GRCh38
rs938671918 Chr.5: 55529175-55529175 on Build GRCh38
rs562703168 Chr.5: 55528938-55528938 on Build GRCh38
rs956390733 Chr.5: 55528950-55528950 on Build GRCh38
rs535347291 Chr.5: 55529089-55529089 on Build GRCh38
rs551425986 Chr.5: 55529030-55529030 on Build GRCh38
rs186785252 Chr.5: 55529249-55529249 on Build GRCh38
rs972161892 Chr.5: 55529155-55529155 on Build GRCh38
rs139427788 Chr.5: 55529271-55529272 on Build GRCh38
rs562434072 Chr.5: 55528848-55528848 on Build GRCh38
rs543533025 Chr.5: 55528897-55528897 on Build GRCh38
rs1030718587 Chr.5: 55529298-55529298 on Build GRCh38
rs1057363254 Chr.5: 55529180-55529180 on Build GRCh38
rs34583149 Chr.5: 55529269-55529270 on Build GRCh38
rs948892417 Chr.5: 55529230-55529230 on Build GRCh38
rs114480853 Chr.5: 55528902-55528902 on Build GRCh38
rs767680476 Chr.5: 55529065-55529065 on Build GRCh38
rs530509541 Chr.5: 55528994-55528994 on Build GRCh38
rs185216251 Chr.5: 55529032-55529032 on Build GRCh38
rs986018109 Chr.5: 55529058-55529059 on Build GRCh38
rs919231397 Chr.5: 55529174-55529174 on Build GRCh38
rs539556942 Chr.5: 55529248-55529248 on Build GRCh38
rs975940958 Chr.5: 55528956-55528956 on Build GRCh38
rs181001784 Chr.5: 55528981-55528981 on Build GRCh38
rs903581290 Chr.5: 55529251-55529252 on Build GRCh38
rs750698301 Chr.5: 55529019-55529019 on Build GRCh38
rs568384023 Chr.5: 55529083-55529083 on Build GRCh38
rs181141579 Chr.5: 55529108-55529108 on Build GRCh38
rs572829632 Chr.5: 55529255-55529255 on Build GRCh38
rs894614426 Chr.5: 55528826-55528826 on Build GRCh38
rs909276985 Chr.5: 55529081-55529081 on Build GRCh38
rs1057403017 Chr.5: 55528819-55528819 on Build GRCh38
rs1003104762 Chr.5: 55528795-55528795 on Build GRCh38
rs1023803040 Chr.5: 55528877-55528883 on Build GRCh38
rs923196851 Chr.5: 55529013-55529013 on Build GRCh38
rs1000924621 Chr.5: 55528926-55528926 on Build GRCh38
rs190320060 Chr.5: 55529082-55529082 on Build GRCh38
rs397738146 Chr.5: 55529281-55529282 on Build GRCh38

View Full Product Details