Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 105099372-105099931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB9BP1
Entrez Gene ID
9366
Gene Name
RAB9B, member RAS oncogene family pseudogene 1
Gene Aliases
RAB9P1
UniGene
Hs.158296
Gene Chromosome Location
Chr.5: 105099474-105100098 on Build GRCh38
Transcripts
NR_000039.2

SNP Detail

SNP ID Chromosome Location
rs879859601 Chr.5: 105099423-105099423 on Build GRCh38
rs4476696 Chr.5: 105099447-105099447 on Build GRCh38
rs890642317 Chr.5: 105099524-105099524 on Build GRCh38
rs951098918 Chr.5: 105099630-105099630 on Build GRCh38
rs936655419 Chr.5: 105099784-105099784 on Build GRCh38
rs745871401 Chr.5: 105099603-105099603 on Build GRCh38
rs553186467 Chr.5: 105099814-105099814 on Build GRCh38
rs565868782 Chr.5: 105099646-105099646 on Build GRCh38
rs981085153 Chr.5: 105099717-105099717 on Build GRCh38
rs146533723 Chr.5: 105099813-105099813 on Build GRCh38
rs761493466 Chr.5: 105099902-105099902 on Build GRCh38
rs534911168 Chr.5: 105099439-105099439 on Build GRCh38
rs4571441 Chr.5: 105099675-105099675 on Build GRCh38
rs554567220 Chr.5: 105099624-105099624 on Build GRCh38
rs573243211 Chr.5: 105099551-105099551 on Build GRCh38
rs112353592 Chr.5: 105099797-105099797 on Build GRCh38
rs951213486 Chr.5: 105099486-105099486 on Build GRCh38
rs989444114 Chr.5: 105099787-105099787 on Build GRCh38
rs973389357 Chr.5: 105099441-105099441 on Build GRCh38
rs555557344 Chr.5: 105099414-105099414 on Build GRCh38
rs192568215 Chr.5: 105099517-105099517 on Build GRCh38
rs571419725 Chr.5: 105099723-105099723 on Build GRCh38
rs945447892 Chr.5: 105099509-105099509 on Build GRCh38
rs184447296 Chr.5: 105099901-105099901 on Build GRCh38
rs549244157 Chr.5: 105099635-105099635 on Build GRCh38
rs370709411 Chr.5: 105099396-105099396 on Build GRCh38
rs913912607 Chr.5: 105099795-105099795 on Build GRCh38
rs148531807 Chr.5: 105099536-105099536 on Build GRCh38
rs748269232 Chr.5: 105099456-105099456 on Build GRCh38
rs530089782 Chr.5: 105099625-105099625 on Build GRCh38
rs535654178 Chr.5: 105099398-105099398 on Build GRCh38
rs577547291 Chr.5: 105099469-105099469 on Build GRCh38
rs953415280 Chr.5: 105099475-105099475 on Build GRCh38
rs995171806 Chr.5: 105099590-105099590 on Build GRCh38
rs769392662 Chr.5: 105099558-105099559 on Build GRCh38
rs142876485 Chr.5: 105099811-105099811 on Build GRCh38
rs878982321 Chr.5: 105099482-105099482 on Build GRCh38
rs890601325 Chr.5: 105099879-105099879 on Build GRCh38
rs565980036 Chr.5: 105099434-105099434 on Build GRCh38
rs188741095 Chr.5: 105099621-105099621 on Build GRCh38
rs537062033 Chr.5: 105099745-105099745 on Build GRCh38
rs543209502 Chr.5: 105099523-105099523 on Build GRCh38
rs184856961 Chr.5: 105099554-105099554 on Build GRCh38
rs181452503 Chr.5: 105099887-105099887 on Build GRCh38
rs557038797 Chr.5: 105099479-105099479 on Build GRCh38
rs113754985 Chr.5: 105099570-105099570 on Build GRCh38
rs552576861 Chr.5: 105099832-105099832 on Build GRCh38
rs1014829669 Chr.5: 105099400-105099400 on Build GRCh38
rs564964702 Chr.5: 105099561-105099561 on Build GRCh38
rs534946537 Chr.5: 105099649-105099649 on Build GRCh38
rs1025203288 Chr.5: 105099665-105099665 on Build GRCh38
rs556638665 Chr.5: 105099782-105099782 on Build GRCh38
rs780225726 Chr.5: 105099732-105099732 on Build GRCh38
rs982562608 Chr.5: 105099653-105099653 on Build GRCh38
rs536616860 Chr.5: 105099478-105099478 on Build GRCh38
rs573727244 Chr.5: 105099798-105099798 on Build GRCh38

View Full Product Details