Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7986562-7987105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S5-TXNDC5
Entrez Gene ID
100526836
Gene Name
BLOC1S5-TXNDC5 readthrough (NMD candidate)
Gene Aliases
MUTED-TXNDC5
UniGene
Hs.150837
Gene Chromosome Location
Chr.6: 7881250-8064414 on Build GRCh38
Transcripts
NR_037616.1 NR_027712.1

Gene Symbol
PIP5K1P1
Entrez Gene ID
206426
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 pseudogene 1
Gene Aliases
-
UniGene
Hs.676966
Gene Chromosome Location
Chr.6: 7986102-7990344 on Build GRCh38
Transcripts
NR_037616.1 NR_027712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611804 c.292C>T p.Q98* Chr.6: 7986828-7986828 on Build GRCh38
1294343 c.89C>G p.S30C Chr.6: 7986625-7986625 on Build GRCh38
1262229 c.160C>T p.R54* Chr.6: 7986696-7986696 on Build GRCh38
6611807 c.159T>C p.H53H Chr.6: 7986695-7986695 on Build GRCh38
5606912 c.381G>A p.K127K Chr.6: 7986917-7986917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767268859 Chr.6: 7987067-7987067 on Build GRCh38
rs766814682 Chr.6: 7986632-7986632 on Build GRCh38
rs768162606 Chr.6: 7986785-7986785 on Build GRCh38
rs763030844 Chr.6: 7987061-7987061 on Build GRCh38
rs768409523 Chr.6: 7986898-7986898 on Build GRCh38
rs779865008 Chr.6: 7986871-7986871 on Build GRCh38
rs777679158 Chr.6: 7986654-7986654 on Build GRCh38
rs778491537 Chr.6: 7987017-7987018 on Build GRCh38
rs768018752 Chr.6: 7986827-7986827 on Build GRCh38
rs756143302 Chr.6: 7986765-7986765 on Build GRCh38
rs772802360 Chr.6: 7986928-7986928 on Build GRCh38
rs751451738 Chr.6: 7986985-7986985 on Build GRCh38
rs748318868 Chr.6: 7986646-7986646 on Build GRCh38
rs571829674 Chr.6: 7986847-7986847 on Build GRCh38
rs768474798 Chr.6: 7986975-7986975 on Build GRCh38
rs943241102 Chr.6: 7986748-7986748 on Build GRCh38
rs772765308 Chr.6: 7987007-7987007 on Build GRCh38
rs764218547 Chr.6: 7986846-7986846 on Build GRCh38
rs769457208 Chr.6: 7986927-7986927 on Build GRCh38
rs776269102 Chr.6: 7986805-7986805 on Build GRCh38
rs757728231 Chr.6: 7986742-7986744 on Build GRCh38
rs548560558 Chr.6: 7986612-7986612 on Build GRCh38
rs766789540 Chr.6: 7986703-7986703 on Build GRCh38
rs769549940 Chr.6: 7986996-7986996 on Build GRCh38
rs750380722 Chr.6: 7986858-7986858 on Build GRCh38
rs758286640 Chr.6: 7986828-7986828 on Build GRCh38
rs745748997 Chr.6: 7986758-7986758 on Build GRCh38
rs780717163 Chr.6: 7986902-7986902 on Build GRCh38
rs747779461 Chr.6: 7987083-7987084 on Build GRCh38
rs76720430 Chr.6: 7986593-7986593 on Build GRCh38
rs763555412 Chr.6: 7986701-7986701 on Build GRCh38
rs749272319 Chr.6: 7986663-7986663 on Build GRCh38
rs763682369 Chr.6: 7986696-7986696 on Build GRCh38
rs781546635 Chr.6: 7986644-7986644 on Build GRCh38
rs575158635 Chr.6: 7986773-7986773 on Build GRCh38
rs756245492 Chr.6: 7986722-7986722 on Build GRCh38
rs780997630 Chr.6: 7986990-7986990 on Build GRCh38
rs751932722 Chr.6: 7986636-7986637 on Build GRCh38
rs762736796 Chr.6: 7986600-7986600 on Build GRCh38
rs747766900 Chr.6: 7986926-7986926 on Build GRCh38
rs907492436 Chr.6: 7986946-7986946 on Build GRCh38
rs754732540 Chr.6: 7986986-7986986 on Build GRCh38
rs753003799 Chr.6: 7986710-7986710 on Build GRCh38
rs764463340 Chr.6: 7986960-7986960 on Build GRCh38
rs765348203 Chr.6: 7986967-7986967 on Build GRCh38
rs754893059 Chr.6: 7987066-7987066 on Build GRCh38
rs754507594 Chr.6: 7986965-7986968 on Build GRCh38
rs1031676371 Chr.6: 7986961-7986961 on Build GRCh38
rs770754768 Chr.6: 7986666-7986666 on Build GRCh38
rs746774973 Chr.6: 7986887-7986887 on Build GRCh38
rs771770100 Chr.6: 7986678-7986678 on Build GRCh38
rs977471152 Chr.6: 7986638-7986638 on Build GRCh38
rs773971036 Chr.6: 7986611-7986611 on Build GRCh38
rs752026751 Chr.6: 7986704-7986704 on Build GRCh38
rs755315008 Chr.6: 7986643-7986643 on Build GRCh38
rs763445648 Chr.6: 7986621-7986621 on Build GRCh38
rs767702799 Chr.6: 7986692-7986692 on Build GRCh38
rs767922879 Chr.6: 7986709-7986709 on Build GRCh38
rs751938918 Chr.6: 7986635-7986635 on Build GRCh38
rs748232621 Chr.6: 7986594-7986594 on Build GRCh38
rs776067970 Chr.6: 7986940-7986940 on Build GRCh38
rs777469784 Chr.6: 7986999-7986999 on Build GRCh38
rs187189984 Chr.6: 7987058-7987058 on Build GRCh38
rs757299468 Chr.6: 7986850-7986850 on Build GRCh38
rs768011044 Chr.6: 7986895-7986897 on Build GRCh38
rs753294808 Chr.6: 7986935-7986936 on Build GRCh38
rs746754876 Chr.6: 7986894-7986894 on Build GRCh38
rs761179580 Chr.6: 7987021-7987021 on Build GRCh38
rs957518186 Chr.6: 7986680-7986680 on Build GRCh38
rs371356877 Chr.6: 7986626-7986626 on Build GRCh38
rs769906323 Chr.6: 7986599-7986599 on Build GRCh38
rs564983624 Chr.6: 7986650-7986650 on Build GRCh38
rs765440437 Chr.6: 7987050-7987050 on Build GRCh38
rs757288518 Chr.6: 7986756-7986756 on Build GRCh38
rs778898152 Chr.6: 7986853-7986853 on Build GRCh38
rs772679670 Chr.6: 7986939-7986939 on Build GRCh38
rs1047425449 Chr.6: 7986827-7986827 on Build GRCh38
rs768976790 Chr.6: 7986584-7986584 on Build GRCh38
rs777960770 Chr.6: 7986746-7986746 on Build GRCh38
rs999252255 Chr.6: 7986949-7986949 on Build GRCh38
rs767128341 Chr.6: 7986613-7986613 on Build GRCh38
rs771884410 Chr.6: 7986763-7986763 on Build GRCh38
rs775138027 Chr.6: 7986681-7986681 on Build GRCh38
rs775979783 Chr.6: 7986697-7986697 on Build GRCh38
rs762060183 Chr.6: 7987044-7987044 on Build GRCh38
rs148316753 Chr.6: 7987003-7987003 on Build GRCh38
rs764744246 Chr.6: 7986813-7986813 on Build GRCh38
rs764522097 Chr.6: 7986623-7986634 on Build GRCh38
rs197129 Chr.6: 7986614-7986614 on Build GRCh38
rs777030713 Chr.6: 7987039-7987039 on Build GRCh38
rs774243738 Chr.6: 7986671-7986671 on Build GRCh38
rs761001643 Chr.6: 7986837-7986837 on Build GRCh38
rs755901785 Chr.6: 7987071-7987071 on Build GRCh38
rs760934412 Chr.6: 7986942-7986942 on Build GRCh38
rs183963795 Chr.6: 7986688-7986688 on Build GRCh38
rs770795839 Chr.6: 7986602-7986602 on Build GRCh38
rs758307676 Chr.6: 7986864-7986864 on Build GRCh38
rs756226384 Chr.6: 7986648-7986648 on Build GRCh38
rs749297428 Chr.6: 7986754-7986754 on Build GRCh38
rs1024231574 Chr.6: 7986619-7986619 on Build GRCh38
rs760172259 Chr.6: 7986929-7986929 on Build GRCh38
rs778809797 Chr.6: 7986757-7986757 on Build GRCh38
rs750337370 Chr.6: 7986819-7986819 on Build GRCh38
rs755402892 Chr.6: 7986706-7986706 on Build GRCh38
rs760165324 Chr.6: 7986620-7986620 on Build GRCh38
rs746724694 Chr.6: 7986772-7986772 on Build GRCh38
rs762044363 Chr.6: 7986962-7986962 on Build GRCh38
rs750970834 Chr.6: 7986624-7986624 on Build GRCh38
rs750470228 Chr.6: 7986971-7986971 on Build GRCh38
rs752861874 Chr.6: 7986752-7986752 on Build GRCh38
rs766375745 Chr.6: 7987064-7987064 on Build GRCh38
rs761210923 Chr.6: 7986807-7986807 on Build GRCh38
rs775875217 Chr.6: 7987009-7987009 on Build GRCh38
rs778964136 Chr.6: 7986814-7986814 on Build GRCh38

View Full Product Details