Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118502500-118503063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD7P3
Entrez Gene ID
23629
Gene Name
bromodomain containing 7 pseudogene 3
Gene Aliases
BP75
UniGene
Hs.648050
Gene Chromosome Location
Chr.6: 118501373-118503846 on Build GRCh38
Transcripts
NR_002730.2 NM_001042475.2 NM_001178035.1 NM_206921.2 XM_005266970.1 XM_005266971.1 XM_005266972.4 XM_006715475.3 XM_011535809.2 XM_011535810.2 XM_011535811.1 XM_017010846.1 XM_017010847.1

Gene Symbol
CEP85L
Entrez Gene ID
387119
Gene Name
centrosomal protein 85 like
Gene Aliases
C6orf204, NY-BR-15, bA57K17.2
UniGene
Hs.656959
Gene Chromosome Location
Chr.6: 118460772-118710101 on Build GRCh38
Transcripts
NR_002730.2 NM_001042475.2 NM_001178035.1 NM_206921.2 XM_005266970.1 XM_005266971.1 XM_005266972.4 XM_006715475.3 XM_011535809.2 XM_011535810.2 XM_011535811.1 XM_017010846.1 XM_017010847.1

SNP Detail

SNP ID Chromosome Location
rs555600272 Chr.6: 118502915-118502915 on Build GRCh38
rs75283875 Chr.6: 118502910-118502910 on Build GRCh38
rs369555078 Chr.6: 118502773-118502773 on Build GRCh38
rs533205182 Chr.6: 118502656-118502656 on Build GRCh38
rs139975871 Chr.6: 118502898-118502898 on Build GRCh38
rs906450926 Chr.6: 118502960-118502960 on Build GRCh38
rs922844868 Chr.6: 118502830-118502830 on Build GRCh38
rs570409654 Chr.6: 118502837-118502837 on Build GRCh38
rs952406551 Chr.6: 118502992-118502992 on Build GRCh38
rs963723613 Chr.6: 118503026-118503026 on Build GRCh38
rs187396423 Chr.6: 118502807-118502807 on Build GRCh38
rs771120681 Chr.6: 118503012-118503012 on Build GRCh38
rs57182451 Chr.6: 118502672-118502672 on Build GRCh38
rs942975291 Chr.6: 118502529-118502529 on Build GRCh38
rs983813805 Chr.6: 118502764-118502764 on Build GRCh38
rs137916843 Chr.6: 118502577-118502577 on Build GRCh38
rs1042124890 Chr.6: 118502852-118502852 on Build GRCh38
rs994073143 Chr.6: 118502962-118502962 on Build GRCh38
rs192325039 Chr.6: 118502958-118502958 on Build GRCh38
rs922870371 Chr.6: 118502541-118502541 on Build GRCh38
rs970668633 Chr.6: 118502659-118502659 on Build GRCh38
rs375323299 Chr.6: 118502825-118502825 on Build GRCh38
rs35225683 Chr.6: 118502801-118502801 on Build GRCh38
rs775346811 Chr.6: 118502987-118502987 on Build GRCh38
rs183117406 Chr.6: 118502772-118502772 on Build GRCh38
rs559096833 Chr.6: 118502572-118502572 on Build GRCh38
rs565165053 Chr.6: 118502752-118502752 on Build GRCh38
rs9481819 Chr.6: 118502638-118502638 on Build GRCh38
rs765156670 Chr.6: 118502582-118502582 on Build GRCh38
rs17339780 Chr.6: 118502612-118502612 on Build GRCh38
rs1025277081 Chr.6: 118502749-118502749 on Build GRCh38
rs190166247 Chr.6: 118502585-118502585 on Build GRCh38
rs575451090 Chr.6: 118502618-118502618 on Build GRCh38
rs1012074991 Chr.6: 118502644-118502644 on Build GRCh38
rs925952726 Chr.6: 118502856-118502856 on Build GRCh38
rs575392883 Chr.6: 118502943-118502943 on Build GRCh38
rs535549775 Chr.6: 118502911-118502911 on Build GRCh38
rs796165964 Chr.6: 118502809-118502809 on Build GRCh38
rs560186671 Chr.6: 118502751-118502751 on Build GRCh38
rs779548488 Chr.6: 118502536-118502536 on Build GRCh38
rs946076533 Chr.6: 118502542-118502542 on Build GRCh38
rs145922924 Chr.6: 118502758-118502758 on Build GRCh38
rs11967375 Chr.6: 118502812-118502812 on Build GRCh38
rs564251422 Chr.6: 118502986-118502986 on Build GRCh38
rs1055917225 Chr.6: 118502882-118502884 on Build GRCh38
rs577891561 Chr.6: 118503008-118503008 on Build GRCh38
rs573051470 Chr.6: 118502583-118502583 on Build GRCh38
rs1024931307 Chr.6: 118502649-118502649 on Build GRCh38
rs73766526 Chr.6: 118502870-118502870 on Build GRCh38
rs564497824 Chr.6: 118502643-118502643 on Build GRCh38

View Full Product Details