Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160466932-160467481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPAL2
Entrez Gene ID
80350
Gene Name
lipoprotein(a) like 2, pseudogene
Gene Aliases
APOA2, APOAL, APOARGC, apo(a)rg-C
UniGene
Hs.654503
Gene Chromosome Location
Chr.6: 160466555-160511124 on Build GRCh38
Transcripts
NR_028092.1 NR_028093.1

SNP Detail

SNP ID Chromosome Location
rs746727711 Chr.6: 160467082-160467082 on Build GRCh38
rs1000826145 Chr.6: 160467419-160467419 on Build GRCh38
rs777919246 Chr.6: 160467140-160467140 on Build GRCh38
rs1021547177 Chr.6: 160466983-160466983 on Build GRCh38
rs55873330 Chr.6: 160467052-160467052 on Build GRCh38
rs991280896 Chr.6: 160467401-160467401 on Build GRCh38
rs559127732 Chr.6: 160467062-160467062 on Build GRCh38
rs200344023 Chr.6: 160467046-160467046 on Build GRCh38
rs985863048 Chr.6: 160467242-160467242 on Build GRCh38
rs115814836 Chr.6: 160467394-160467394 on Build GRCh38
rs575634438 Chr.6: 160467014-160467014 on Build GRCh38
rs919190897 Chr.6: 160467351-160467351 on Build GRCh38
rs558988409 Chr.6: 160467012-160467012 on Build GRCh38
rs1011885145 Chr.6: 160466959-160466959 on Build GRCh38
rs967999593 Chr.6: 160467200-160467200 on Build GRCh38
rs1034494385 Chr.6: 160467163-160467163 on Build GRCh38
rs954556813 Chr.6: 160467231-160467231 on Build GRCh38
rs192112751 Chr.6: 160466987-160466987 on Build GRCh38
rs878924692 Chr.6: 160467023-160467023 on Build GRCh38
rs551641295 Chr.6: 160467441-160467442 on Build GRCh38
rs893760234 Chr.6: 160467124-160467124 on Build GRCh38
rs200204404 Chr.6: 160467008-160467008 on Build GRCh38
rs373634678 Chr.6: 160467447-160467447 on Build GRCh38
rs758339064 Chr.6: 160467006-160467006 on Build GRCh38
rs202061167 Chr.6: 160467045-160467045 on Build GRCh38
rs372430111 Chr.6: 160467051-160467052 on Build GRCh38
rs71033580 Chr.6: 160467052-160467052 on Build GRCh38
rs985915547 Chr.6: 160467448-160467448 on Build GRCh38
rs993064040 Chr.6: 160467190-160467190 on Build GRCh38
rs770976904 Chr.6: 160467370-160467370 on Build GRCh38
rs1020832100 Chr.6: 160467398-160467398 on Build GRCh38
rs1036312344 Chr.6: 160467349-160467349 on Build GRCh38
rs539329197 Chr.6: 160467329-160467329 on Build GRCh38
rs978958568 Chr.6: 160467016-160467016 on Build GRCh38
rs573535970 Chr.6: 160466988-160466988 on Build GRCh38
rs114991458 Chr.6: 160467128-160467128 on Build GRCh38
rs973084482 Chr.6: 160467283-160467283 on Build GRCh38
rs946831435 Chr.6: 160467424-160467424 on Build GRCh38
rs62441868 Chr.6: 160466989-160466989 on Build GRCh38
rs550324450 Chr.6: 160467134-160467134 on Build GRCh38
rs567115435 Chr.6: 160467271-160467271 on Build GRCh38
rs182218437 Chr.6: 160467440-160467440 on Build GRCh38
rs16891630 Chr.6: 160467074-160467074 on Build GRCh38
rs1001603416 Chr.6: 160467353-160467354 on Build GRCh38
rs1014368972 Chr.6: 160467395-160467395 on Build GRCh38
rs915323986 Chr.6: 160467417-160467417 on Build GRCh38
rs553555139 Chr.6: 160467234-160467234 on Build GRCh38
rs985869971 Chr.6: 160467002-160467002 on Build GRCh38
rs536825221 Chr.6: 160466977-160466977 on Build GRCh38
rs754011662 Chr.6: 160467068-160467068 on Build GRCh38
rs1030106609 Chr.6: 160467142-160467142 on Build GRCh38
rs111544537 Chr.6: 160466957-160466957 on Build GRCh38
rs954441722 Chr.6: 160466991-160466991 on Build GRCh38
rs1055788693 Chr.6: 160467357-160467357 on Build GRCh38
rs567317767 Chr.6: 160467162-160467162 on Build GRCh38
rs111795417 Chr.6: 160467046-160467047 on Build GRCh38
rs184704895 Chr.6: 160467172-160467172 on Build GRCh38
rs1026437681 Chr.6: 160467297-160467297 on Build GRCh38
rs963340090 Chr.6: 160467279-160467279 on Build GRCh38
rs551305509 Chr.6: 160467118-160467118 on Build GRCh38
rs544363809 Chr.6: 160467031-160467031 on Build GRCh38
rs116199835 Chr.6: 160467352-160467352 on Build GRCh38
rs180740118 Chr.6: 160467217-160467217 on Build GRCh38
rs561406027 Chr.6: 160467034-160467034 on Build GRCh38
rs978453290 Chr.6: 160467206-160467206 on Build GRCh38
rs944517431 Chr.6: 160467307-160467307 on Build GRCh38
rs1020533079 Chr.6: 160467197-160467197 on Build GRCh38
rs960186333 Chr.6: 160467175-160467175 on Build GRCh38
rs138394255 Chr.6: 160467353-160467353 on Build GRCh38
rs903099722 Chr.6: 160467138-160467138 on Build GRCh38
rs1017355087 Chr.6: 160467270-160467270 on Build GRCh38
rs950933477 Chr.6: 160467314-160467314 on Build GRCh38
rs869036530 Chr.6: 160467053-160467054 on Build GRCh38
rs1051648471 Chr.6: 160467453-160467453 on Build GRCh38
rs925585066 Chr.6: 160467210-160467210 on Build GRCh38
rs1007237845 Chr.6: 160467259-160467259 on Build GRCh38
rs1042375404 Chr.6: 160467428-160467428 on Build GRCh38
rs963091740 Chr.6: 160467024-160467024 on Build GRCh38
rs530558952 Chr.6: 160467129-160467129 on Build GRCh38

View Full Product Details