Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5121635-5122185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF890P
Entrez Gene ID
645700
Gene Name
zinc finger protein 890, pseudogene
Gene Aliases
-
UniGene
Hs.690815
Gene Chromosome Location
Chr.7: 5121310-5144546 on Build GRCh38
Transcripts
NR_034163.1

SNP Detail

SNP ID Chromosome Location
rs62441986 Chr.7: 5122009-5122009 on Build GRCh38
rs780593761 Chr.7: 5122018-5122018 on Build GRCh38
rs889716708 Chr.7: 5121747-5121747 on Build GRCh38
rs535103343 Chr.7: 5121841-5121841 on Build GRCh38
rs777348267 Chr.7: 5122111-5122111 on Build GRCh38
rs780697989 Chr.7: 5121858-5121858 on Build GRCh38
rs775553899 Chr.7: 5121854-5121854 on Build GRCh38
rs762363613 Chr.7: 5122040-5122041 on Build GRCh38
rs764033888 Chr.7: 5121941-5121941 on Build GRCh38
rs779325912 Chr.7: 5122098-5122098 on Build GRCh38
rs750736690 Chr.7: 5121995-5121995 on Build GRCh38
rs1012268065 Chr.7: 5121800-5121800 on Build GRCh38
rs755485077 Chr.7: 5122047-5122047 on Build GRCh38
rs760449365 Chr.7: 5121900-5121900 on Build GRCh38
rs557502386 Chr.7: 5121958-5121958 on Build GRCh38
rs1023618917 Chr.7: 5121815-5121815 on Build GRCh38
rs573980229 Chr.7: 5122161-5122161 on Build GRCh38
rs751638134 Chr.7: 5121944-5121944 on Build GRCh38
rs929809028 Chr.7: 5122119-5122119 on Build GRCh38
rs757901957 Chr.7: 5122104-5122104 on Build GRCh38
rs747657937 Chr.7: 5121875-5121875 on Build GRCh38
rs539267496 Chr.7: 5121957-5121957 on Build GRCh38
rs778506251 Chr.7: 5121871-5121871 on Build GRCh38
rs1056282952 Chr.7: 5122141-5122141 on Build GRCh38
rs772914465 Chr.7: 5121899-5121899 on Build GRCh38
rs375007473 Chr.7: 5121903-5121903 on Build GRCh38
rs556326100 Chr.7: 5121939-5121939 on Build GRCh38
rs994304870 Chr.7: 5121773-5121773 on Build GRCh38
rs768007477 Chr.7: 5121867-5121867 on Build GRCh38
rs182791013 Chr.7: 5121898-5121898 on Build GRCh38
rs957095529 Chr.7: 5121919-5121919 on Build GRCh38
rs922368348 Chr.7: 5121960-5121960 on Build GRCh38
rs543735583 Chr.7: 5122124-5122124 on Build GRCh38
rs770674202 Chr.7: 5122153-5122153 on Build GRCh38
rs551970333 Chr.7: 5121749-5121749 on Build GRCh38
rs570381699 Chr.7: 5121761-5121761 on Build GRCh38
rs568609002 Chr.7: 5121961-5121961 on Build GRCh38
rs754343026 Chr.7: 5122020-5122020 on Build GRCh38
rs746668173 Chr.7: 5122128-5122128 on Build GRCh38
rs933665250 Chr.7: 5121676-5121676 on Build GRCh38
rs924171891 Chr.7: 5121887-5121887 on Build GRCh38
rs62441978 Chr.7: 5121791-5121791 on Build GRCh38
rs747687301 Chr.7: 5122101-5122101 on Build GRCh38
rs62441979 Chr.7: 5121881-5121881 on Build GRCh38
rs537774310 Chr.7: 5121767-5121767 on Build GRCh38
rs910863264 Chr.7: 5121964-5121964 on Build GRCh38
rs62441980 Chr.7: 5121921-5121921 on Build GRCh38
rs555366018 Chr.7: 5122130-5122130 on Build GRCh38
rs1038693127 Chr.7: 5122028-5122028 on Build GRCh38
rs776486346 Chr.7: 5121928-5121928 on Build GRCh38
rs1049354543 Chr.7: 5121738-5121738 on Build GRCh38
rs1035106764 Chr.7: 5121778-5121778 on Build GRCh38
rs980080453 Chr.7: 5121870-5121870 on Build GRCh38
rs897032629 Chr.7: 5122041-5122041 on Build GRCh38
rs960749267 Chr.7: 5121798-5121798 on Build GRCh38
rs756616200 Chr.7: 5121847-5121847 on Build GRCh38
rs941320352 Chr.7: 5121998-5121998 on Build GRCh38
rs749814640 Chr.7: 5121862-5121862 on Build GRCh38
rs568086420 Chr.7: 5121793-5121793 on Build GRCh38

View Full Product Details