Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5822981-5823560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF815P
Entrez Gene ID
401303
Gene Name
zinc finger protein 815, pseudogene
Gene Aliases
ZNF815
UniGene
Hs.372707
Gene Chromosome Location
Chr.7: 5823160-5854435 on Build GRCh38
Transcripts
NR_023382.1

SNP Detail

SNP ID Chromosome Location
rs561185355 Chr.7: 5823514-5823514 on Build GRCh38
rs1055011200 Chr.7: 5823469-5823469 on Build GRCh38
rs999006953 Chr.7: 5823363-5823363 on Build GRCh38
rs542618762 Chr.7: 5823498-5823498 on Build GRCh38
rs968817735 Chr.7: 5823232-5823232 on Build GRCh38
rs888390680 Chr.7: 5823071-5823071 on Build GRCh38
rs61738473 Chr.7: 5823329-5823329 on Build GRCh38
rs575136274 Chr.7: 5823483-5823483 on Build GRCh38
rs976438532 Chr.7: 5823177-5823177 on Build GRCh38
rs927796084 Chr.7: 5823456-5823456 on Build GRCh38
rs901309345 Chr.7: 5823536-5823536 on Build GRCh38
rs1032373161 Chr.7: 5823234-5823234 on Build GRCh38
rs888250918 Chr.7: 5823187-5823187 on Build GRCh38
rs140624512 Chr.7: 5823174-5823174 on Build GRCh38
rs761896587 Chr.7: 5823358-5823358 on Build GRCh38
rs546782835 Chr.7: 5823236-5823236 on Build GRCh38
rs939265573 Chr.7: 5823335-5823335 on Build GRCh38
rs769926321 Chr.7: 5823408-5823408 on Build GRCh38
rs899579966 Chr.7: 5823053-5823053 on Build GRCh38
rs2287589 Chr.7: 5823017-5823017 on Build GRCh38
rs1041382896 Chr.7: 5823027-5823027 on Build GRCh38
rs977555479 Chr.7: 5823233-5823233 on Build GRCh38
rs753928470 Chr.7: 5823078-5823078 on Build GRCh38
rs947626523 Chr.7: 5823019-5823019 on Build GRCh38
rs998416965 Chr.7: 5823381-5823381 on Build GRCh38
rs757200450 Chr.7: 5823145-5823145 on Build GRCh38
rs961103207 Chr.7: 5823443-5823443 on Build GRCh38
rs939876584 Chr.7: 5823295-5823295 on Build GRCh38
rs567851653 Chr.7: 5823121-5823121 on Build GRCh38
rs973002422 Chr.7: 5823449-5823449 on Build GRCh38
rs958390759 Chr.7: 5823472-5823472 on Build GRCh38
rs1036859779 Chr.7: 5823310-5823310 on Build GRCh38
rs1048247626 Chr.7: 5823057-5823057 on Build GRCh38
rs552094916 Chr.7: 5823184-5823185 on Build GRCh38
rs906448848 Chr.7: 5823204-5823204 on Build GRCh38
rs552665471 Chr.7: 5823117-5823117 on Build GRCh38
rs1002961110 Chr.7: 5823098-5823098 on Build GRCh38
rs373998959 Chr.7: 5823028-5823028 on Build GRCh38
rs1021730283 Chr.7: 5823116-5823116 on Build GRCh38
rs1012926412 Chr.7: 5823228-5823228 on Build GRCh38
rs373985499 Chr.7: 5823467-5823467 on Build GRCh38
rs1043024809 Chr.7: 5823474-5823474 on Build GRCh38
rs983839883 Chr.7: 5823282-5823282 on Build GRCh38
rs909721607 Chr.7: 5823286-5823286 on Build GRCh38
rs538343776 Chr.7: 5823457-5823457 on Build GRCh38
rs920913210 Chr.7: 5823256-5823257 on Build GRCh38
rs977609544 Chr.7: 5823133-5823133 on Build GRCh38
rs904607163 Chr.7: 5823360-5823360 on Build GRCh38
rs187642415 Chr.7: 5823144-5823144 on Build GRCh38
rs897784469 Chr.7: 5823096-5823096 on Build GRCh38
rs991646238 Chr.7: 5823107-5823107 on Build GRCh38
rs61734878 Chr.7: 5823231-5823231 on Build GRCh38
rs540398872 Chr.7: 5823109-5823109 on Build GRCh38
rs185011379 Chr.7: 5823355-5823355 on Build GRCh38
rs948755080 Chr.7: 5823470-5823470 on Build GRCh38
rs997119907 Chr.7: 5823055-5823055 on Build GRCh38
rs542569073 Chr.7: 5823182-5823182 on Build GRCh38
rs1003321188 Chr.7: 5823207-5823207 on Build GRCh38
rs780592012 Chr.7: 5823400-5823400 on Build GRCh38
rs752340654 Chr.7: 5823410-5823410 on Build GRCh38
rs904638075 Chr.7: 5823496-5823496 on Build GRCh38
rs541285035 Chr.7: 5823345-5823345 on Build GRCh38
rs569166915 Chr.7: 5823268-5823268 on Build GRCh38
rs78789621 Chr.7: 5823099-5823099 on Build GRCh38
rs943811522 Chr.7: 5823030-5823030 on Build GRCh38
rs386709777 Chr.7: 5823343-5823345 on Build GRCh38
rs571804331 Chr.7: 5823241-5823241 on Build GRCh38
rs115283925 Chr.7: 5823003-5823003 on Build GRCh38
rs1048278525 Chr.7: 5823186-5823186 on Build GRCh38
rs144087882 Chr.7: 5823250-5823250 on Build GRCh38
rs1035683577 Chr.7: 5823209-5823209 on Build GRCh38
rs991669459 Chr.7: 5823009-5823009 on Build GRCh38
rs894576121 Chr.7: 5823220-5823220 on Build GRCh38
rs113472831 Chr.7: 5823150-5823150 on Build GRCh38
rs556979662 Chr.7: 5823461-5823461 on Build GRCh38
rs991469149 Chr.7: 5823499-5823499 on Build GRCh38
rs534155318 Chr.7: 5823416-5823416 on Build GRCh38
rs779952289 Chr.7: 5823270-5823270 on Build GRCh38
rs62000374 Chr.7: 5823456-5823456 on Build GRCh38
rs755727201 Chr.7: 5823501-5823501 on Build GRCh38
rs909667624 Chr.7: 5823434-5823434 on Build GRCh38
rs551001506 Chr.7: 5823263-5823263 on Build GRCh38
rs958809565 Chr.7: 5823104-5823104 on Build GRCh38
rs1057446309 Chr.7: 5823336-5823336 on Build GRCh38
rs7806927 Chr.7: 5823343-5823343 on Build GRCh38
rs192657530 Chr.7: 5823169-5823169 on Build GRCh38
rs745888700 Chr.7: 5823172-5823172 on Build GRCh38
rs1004106583 Chr.7: 5823075-5823075 on Build GRCh38
rs554801157 Chr.7: 5823041-5823041 on Build GRCh38
rs146475829 Chr.7: 5823428-5823428 on Build GRCh38
rs1036909850 Chr.7: 5823200-5823200 on Build GRCh38
rs7784409 Chr.7: 5823199-5823199 on Build GRCh38
rs538403720 Chr.7: 5823522-5823522 on Build GRCh38
rs550605200 Chr.7: 5823368-5823368 on Build GRCh38
rs540135203 Chr.7: 5823166-5823167 on Build GRCh38
rs1041153893 Chr.7: 5823050-5823050 on Build GRCh38

View Full Product Details