Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75415274-75415822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSUN5P1
Entrez Gene ID
155400
Gene Name
NOP2/Sun RNA methyltransferase family member 5 pseudogene 1
Gene Aliases
NSUN5B, WBSCR20B
UniGene
Hs.436034
Gene Chromosome Location
Chr.7: 75410322-75416792 on Build GRCh38
Transcripts
NR_033322.3 NR_104013.1

SNP Detail

SNP ID Chromosome Location
rs782305558 Chr.7: 75415634-75415634 on Build GRCh38
rs782672372 Chr.7: 75415671-75415671 on Build GRCh38
rs782766741 Chr.7: 75415725-75415725 on Build GRCh38
rs782631217 Chr.7: 75415686-75415686 on Build GRCh38
rs782403622 Chr.7: 75415700-75415700 on Build GRCh38
rs782289096 Chr.7: 75415699-75415699 on Build GRCh38
rs587734679 Chr.7: 75415304-75415304 on Build GRCh38
rs587677405 Chr.7: 75415719-75415719 on Build GRCh38
rs587702919 Chr.7: 75415548-75415548 on Build GRCh38
rs141502909 Chr.7: 75415341-75415341 on Build GRCh38
rs782710129 Chr.7: 75415657-75415657 on Build GRCh38
rs782571506 Chr.7: 75415624-75415624 on Build GRCh38
rs782179452 Chr.7: 75415630-75415630 on Build GRCh38
rs148778056 Chr.7: 75415300-75415300 on Build GRCh38
rs782004267 Chr.7: 75415639-75415639 on Build GRCh38
rs782511967 Chr.7: 75415611-75415611 on Build GRCh38
rs587612047 Chr.7: 75415642-75415642 on Build GRCh38
rs11768785 Chr.7: 75415544-75415544 on Build GRCh38
rs782142525 Chr.7: 75415721-75415721 on Build GRCh38
rs782429322 Chr.7: 75415602-75415602 on Build GRCh38
rs781799970 Chr.7: 75415601-75415601 on Build GRCh38
rs188183330 Chr.7: 75415472-75415472 on Build GRCh38
rs782150077 Chr.7: 75415652-75415652 on Build GRCh38
rs782273000 Chr.7: 75415673-75415673 on Build GRCh38
rs587707577 Chr.7: 75415635-75415635 on Build GRCh38
rs587666548 Chr.7: 75415483-75415483 on Build GRCh38
rs1823616 Chr.7: 75415352-75415352 on Build GRCh38
rs762484293 Chr.7: 75415347-75415347 on Build GRCh38
rs781845144 Chr.7: 75415726-75415726 on Build GRCh38
rs782572455 Chr.7: 75415731-75415731 on Build GRCh38
rs782264209 Chr.7: 75415769-75415769 on Build GRCh38
rs782203440 Chr.7: 75415687-75415687 on Build GRCh38
rs199616834 Chr.7: 75415788-75415788 on Build GRCh38
rs587600064 Chr.7: 75415682-75415682 on Build GRCh38
rs587741178 Chr.7: 75415416-75415416 on Build GRCh38
rs782357390 Chr.7: 75415706-75415706 on Build GRCh38
rs782459530 Chr.7: 75415623-75415623 on Build GRCh38
rs781949108 Chr.7: 75415643-75415643 on Build GRCh38
rs587639170 Chr.7: 75415644-75415644 on Build GRCh38
rs781941981 Chr.7: 75415717-75415717 on Build GRCh38
rs781912256 Chr.7: 75415691-75415691 on Build GRCh38
rs587638127 Chr.7: 75415558-75415558 on Build GRCh38
rs782811397 Chr.7: 75415604-75415604 on Build GRCh38
rs200393397 Chr.7: 75415654-75415654 on Build GRCh38
rs782033510 Chr.7: 75415650-75415650 on Build GRCh38
rs782114418 Chr.7: 75415705-75415705 on Build GRCh38
rs763787213 Chr.7: 75415348-75415348 on Build GRCh38
rs781791104 Chr.7: 75415661-75415661 on Build GRCh38
rs139385548 Chr.7: 75415473-75415473 on Build GRCh38
rs191457015 Chr.7: 75415480-75415480 on Build GRCh38
rs782468230 Chr.7: 75415730-75415730 on Build GRCh38
rs782050198 Chr.7: 75415718-75415718 on Build GRCh38
rs782510452 Chr.7: 75415674-75415674 on Build GRCh38
rs587611396 Chr.7: 75415459-75415459 on Build GRCh38
rs587712205 Chr.7: 75415748-75415748 on Build GRCh38
rs781887175 Chr.7: 75415605-75415605 on Build GRCh38
rs781783926 Chr.7: 75415741-75415741 on Build GRCh38
rs782314813 Chr.7: 75415789-75415789 on Build GRCh38
rs782555583 Chr.7: 75415662-75415662 on Build GRCh38
rs587605563 Chr.7: 75415768-75415768 on Build GRCh38
rs201218459 Chr.7: 75415655-75415655 on Build GRCh38
rs750265652 Chr.7: 75415467-75415467 on Build GRCh38
rs781887554 Chr.7: 75415720-75415720 on Build GRCh38
rs11768063 Chr.7: 75415413-75415413 on Build GRCh38
rs236651 Chr.7: 75415552-75415552 on Build GRCh38
rs782617786 Chr.7: 75415785-75415785 on Build GRCh38
rs782639230 Chr.7: 75415619-75415619 on Build GRCh38
rs587683335 Chr.7: 75415408-75415408 on Build GRCh38

View Full Product Details