Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75418567-75419112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POM121C
Entrez Gene ID
100101267
Gene Name
POM121 transmembrane nucleoporin C
Gene Aliases
POM121-2
UniGene
Hs.712563
Gene Chromosome Location
Chr.7: 75416781-75486288 on Build GRCh38
Transcripts
NM_001099415.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4827745 c.2939G>A p.R980Q Chr.7: 75418821-75418821 on Build GRCh38
5167247 c.2959A>C p.K987Q Chr.7: 75418801-75418801 on Build GRCh38
421882 c.2889C>G p.S963S Chr.7: 75418871-75418871 on Build GRCh38
1131524 c.2897C>T p.A966V Chr.7: 75418863-75418863 on Build GRCh38
6349284 c.2912C>T p.P971L Chr.7: 75418848-75418848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587685002 Chr.7: 75418769-75418769 on Build GRCh38
rs587634020 Chr.7: 75418707-75418707 on Build GRCh38
rs2429587 Chr.7: 75419051-75419051 on Build GRCh38
rs375388422 Chr.7: 75418849-75418849 on Build GRCh38
rs782068741 Chr.7: 75418839-75418839 on Build GRCh38
rs116287082 Chr.7: 75419037-75419037 on Build GRCh38
rs587652804 Chr.7: 75418942-75418942 on Build GRCh38
rs373682478 Chr.7: 75418901-75418901 on Build GRCh38
rs58148454 Chr.7: 75418692-75418692 on Build GRCh38
rs782015839 Chr.7: 75418813-75418813 on Build GRCh38
rs782314190 Chr.7: 75419066-75419067 on Build GRCh38
rs1128186 Chr.7: 75418881-75418881 on Build GRCh38
rs782790642 Chr.7: 75418908-75418908 on Build GRCh38
rs2429586 Chr.7: 75419043-75419043 on Build GRCh38
rs782300846 Chr.7: 75418886-75418886 on Build GRCh38
rs185328171 Chr.7: 75418848-75418848 on Build GRCh38
rs236656 Chr.7: 75419035-75419035 on Build GRCh38
rs782592676 Chr.7: 75418871-75418871 on Build GRCh38
rs781827255 Chr.7: 75418833-75418833 on Build GRCh38
rs782391614 Chr.7: 75418890-75418890 on Build GRCh38
rs587773219 Chr.7: 75418692-75418692 on Build GRCh38
rs782163319 Chr.7: 75418903-75418903 on Build GRCh38
rs2533486 Chr.7: 75418945-75418945 on Build GRCh38
rs587602776 Chr.7: 75419016-75419016 on Build GRCh38
rs781924090 Chr.7: 75418902-75418902 on Build GRCh38
rs781980398 Chr.7: 75418895-75418895 on Build GRCh38
rs236655 Chr.7: 75418889-75418889 on Build GRCh38
rs782193402 Chr.7: 75418872-75418872 on Build GRCh38
rs781928808 Chr.7: 75418787-75418787 on Build GRCh38
rs367897445 Chr.7: 75418863-75418863 on Build GRCh38
rs367704117 Chr.7: 75418865-75418865 on Build GRCh38
rs376495903 Chr.7: 75418661-75418661 on Build GRCh38
rs370125686 Chr.7: 75418887-75418887 on Build GRCh38
rs200953843 Chr.7: 75418807-75418807 on Build GRCh38
rs782708906 Chr.7: 75418847-75418847 on Build GRCh38
rs782763551 Chr.7: 75418830-75418830 on Build GRCh38
rs587769635 Chr.7: 75419088-75419088 on Build GRCh38
rs782045130 Chr.7: 75418806-75418806 on Build GRCh38
rs2429585 Chr.7: 75419022-75419022 on Build GRCh38
rs782132881 Chr.7: 75418829-75418829 on Build GRCh38
rs113561610 Chr.7: 75418962-75418962 on Build GRCh38
rs782496505 Chr.7: 75418869-75418869 on Build GRCh38

View Full Product Details