Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112501123-112501486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTL
Entrez Gene ID
150465
Gene Name
tubulin tyrosine ligase
Gene Aliases
-
UniGene
Hs.358997
Gene Chromosome Location
Chr.2: 112482127-112532646 on Build GRCh38
Transcripts
NM_153712.4 XM_005263599.3 XM_011510665.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3797848 c.509T>C p.L170P Chr.2: 112501245-112501245 on Build GRCh38
6370170 c.590G>A p.R197H Chr.2: 112501326-112501326 on Build GRCh38
6337188 c.533A>C p.Q178P Chr.2: 112501269-112501269 on Build GRCh38
6247759 c.566C>A p.P189H Chr.2: 112501302-112501302 on Build GRCh38
4953910 c.559G>T p.E187* Chr.2: 112501295-112501295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023867279 Chr.2: 112501443-112501443 on Build GRCh38
rs969548839 Chr.2: 112501457-112501457 on Build GRCh38
rs763593707 Chr.2: 112501267-112501267 on Build GRCh38
rs778614090 Chr.2: 112501306-112501306 on Build GRCh38
rs1047500880 Chr.2: 112501222-112501222 on Build GRCh38
rs778898752 Chr.2: 112501192-112501192 on Build GRCh38
rs756993339 Chr.2: 112501327-112501327 on Build GRCh38
rs755697005 Chr.2: 112501276-112501276 on Build GRCh38
rs745710997 Chr.2: 112501313-112501313 on Build GRCh38
rs751543163 Chr.2: 112501320-112501320 on Build GRCh38
rs750915562 Chr.2: 112501265-112501265 on Build GRCh38
rs780894420 Chr.2: 112501172-112501172 on Build GRCh38
rs368240263 Chr.2: 112501219-112501219 on Build GRCh38
rs754518717 Chr.2: 112501171-112501171 on Build GRCh38
rs752204601 Chr.2: 112501270-112501270 on Build GRCh38
rs760250189 Chr.2: 112501391-112501391 on Build GRCh38
rs767022904 Chr.2: 112501269-112501269 on Build GRCh38
rs777590105 Chr.2: 112501183-112501183 on Build GRCh38
rs574780881 Chr.2: 112501451-112501451 on Build GRCh38
rs765707258 Chr.2: 112501163-112501163 on Build GRCh38
rs1011589010 Chr.2: 112501434-112501434 on Build GRCh38
rs760770201 Chr.2: 112501232-112501232 on Build GRCh38
rs748364962 Chr.2: 112501346-112501346 on Build GRCh38
rs879284795 Chr.2: 112501338-112501339 on Build GRCh38
rs2943625 Chr.2: 112501247-112501247 on Build GRCh38
rs776895890 Chr.2: 112501341-112501341 on Build GRCh38
rs763562520 Chr.2: 112501392-112501392 on Build GRCh38
rs560753574 Chr.2: 112501378-112501378 on Build GRCh38
rs1029369473 Chr.2: 112501340-112501340 on Build GRCh38
rs763800735 Chr.2: 112501279-112501279 on Build GRCh38
rs745843295 Chr.2: 112501196-112501196 on Build GRCh38
rs753493156 Chr.2: 112501295-112501295 on Build GRCh38
rs558637408 Chr.2: 112501146-112501146 on Build GRCh38
rs375362800 Chr.2: 112501191-112501191 on Build GRCh38
rs982244848 Chr.2: 112501459-112501459 on Build GRCh38
rs768811629 Chr.2: 112501246-112501246 on Build GRCh38
rs775002334 Chr.2: 112501380-112501380 on Build GRCh38
rs757054225 Chr.2: 112501296-112501296 on Build GRCh38
rs765574711 Chr.2: 112501253-112501253 on Build GRCh38
rs747027341 Chr.2: 112501326-112501326 on Build GRCh38
rs371924216 Chr.2: 112501190-112501190 on Build GRCh38
rs770160304 Chr.2: 112501358-112501358 on Build GRCh38
rs2918763 Chr.2: 112501339-112501339 on Build GRCh38
rs929970394 Chr.2: 112501211-112501211 on Build GRCh38
rs763432558 Chr.2: 112501373-112501373 on Build GRCh38
rs772109218 Chr.2: 112501197-112501197 on Build GRCh38
rs372561557 Chr.2: 112501371-112501371 on Build GRCh38
rs958344381 Chr.2: 112501400-112501400 on Build GRCh38
rs1033089299 Chr.2: 112501397-112501397 on Build GRCh38
rs755787055 Chr.2: 112501180-112501180 on Build GRCh38
rs762100064 Chr.2: 112501159-112501159 on Build GRCh38
rs752366367 Chr.2: 112501179-112501179 on Build GRCh38
rs1039566594 Chr.2: 112501302-112501302 on Build GRCh38

View Full Product Details