Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112490784-112491139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT2B
Entrez Gene ID
7482
Gene Name
Wnt family member 2B
Gene Aliases
WNT13
UniGene
Hs.258575
Gene Chromosome Location
Chr.1: 112466541-112521288 on Build GRCh38
Transcripts
NM_001291880.1 NM_004185.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4576118 c.81A>C p.V27V Chr.1: 112491037-112491037 on Build GRCh38
2119835 c.105G>A p.A35A Chr.1: 112491061-112491061 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376076633 Chr.1: 112490987-112490987 on Build GRCh38
rs550786359 Chr.1: 112490879-112490879 on Build GRCh38
rs1027468413 Chr.1: 112490959-112490959 on Build GRCh38
rs957113810 Chr.1: 112491031-112491031 on Build GRCh38
rs745780586 Chr.1: 112491010-112491010 on Build GRCh38
rs762577452 Chr.1: 112491027-112491027 on Build GRCh38
rs751612302 Chr.1: 112491026-112491027 on Build GRCh38
rs767306905 Chr.1: 112490894-112490894 on Build GRCh38
rs565227204 Chr.1: 112490858-112490858 on Build GRCh38
rs773932825 Chr.1: 112491051-112491051 on Build GRCh38
rs758607059 Chr.1: 112491088-112491088 on Build GRCh38
rs61820474 Chr.1: 112490837-112490837 on Build GRCh38
rs781283417 Chr.1: 112491004-112491004 on Build GRCh38
rs754275874 Chr.1: 112490976-112490976 on Build GRCh38
rs373335061 Chr.1: 112491005-112491005 on Build GRCh38
rs775031995 Chr.1: 112491012-112491012 on Build GRCh38
rs766689180 Chr.1: 112491062-112491062 on Build GRCh38
rs992249635 Chr.1: 112490826-112490826 on Build GRCh38
rs781553902 Chr.1: 112491113-112491113 on Build GRCh38
rs768337962 Chr.1: 112491042-112491042 on Build GRCh38
rs754363592 Chr.1: 112491067-112491067 on Build GRCh38
rs760065435 Chr.1: 112491081-112491081 on Build GRCh38
rs770796349 Chr.1: 112491003-112491003 on Build GRCh38
rs982636991 Chr.1: 112491029-112491029 on Build GRCh38
rs935667687 Chr.1: 112490886-112490886 on Build GRCh38
rs530817268 Chr.1: 112490862-112490862 on Build GRCh38
rs529862620 Chr.1: 112490973-112490973 on Build GRCh38
rs530977981 Chr.1: 112491066-112491066 on Build GRCh38
rs779001066 Chr.1: 112490986-112490986 on Build GRCh38
rs928055658 Chr.1: 112491030-112491030 on Build GRCh38
rs963065295 Chr.1: 112491087-112491087 on Build GRCh38
rs181521752 Chr.1: 112490848-112490848 on Build GRCh38
rs891511916 Chr.1: 112490911-112490911 on Build GRCh38
rs552143676 Chr.1: 112491081-112491081 on Build GRCh38
rs1018176524 Chr.1: 112491053-112491053 on Build GRCh38
rs778155541 Chr.1: 112491089-112491089 on Build GRCh38
rs761031689 Chr.1: 112491060-112491060 on Build GRCh38
rs565788658 Chr.1: 112491082-112491082 on Build GRCh38
rs1010081196 Chr.1: 112490919-112490919 on Build GRCh38
rs897682876 Chr.1: 112490910-112490910 on Build GRCh38
rs755044291 Chr.1: 112490983-112490983 on Build GRCh38
rs559367749 Chr.1: 112490827-112490827 on Build GRCh38
rs751768135 Chr.1: 112491098-112491098 on Build GRCh38
rs777778243 Chr.1: 112490990-112490990 on Build GRCh38
rs376409195 Chr.1: 112491086-112491086 on Build GRCh38
rs560898908 Chr.1: 112490898-112490898 on Build GRCh38
rs746787857 Chr.1: 112490998-112490998 on Build GRCh38
rs1050886535 Chr.1: 112490972-112490972 on Build GRCh38
rs757153475 Chr.1: 112491110-112491110 on Build GRCh38

View Full Product Details