Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69566289-69566692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H2C
Entrez Gene ID
728340
Gene Name
GTF2H2 family member C
Gene Aliases
GTF2H2C_2
UniGene
Hs.191356
Gene Chromosome Location
Chr.5: 69560136-69592903 on Build GRCh38
Transcripts
NM_001098728.1 XM_005248588.4 XM_005248590.3 XM_005248592.3 XM_006714688.3 XM_011543615.1 XM_017009804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3072313 c.150T>C p.H50H Chr.5: 69566604-69566604 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199709030 Chr.5: 69566615-69566615 on Build GRCh38
rs561259023 Chr.5: 69566405-69566405 on Build GRCh38
rs1039255320 Chr.5: 69566406-69566406 on Build GRCh38
rs1008712958 Chr.5: 69566397-69566397 on Build GRCh38
rs747084183 Chr.5: 69566544-69566545 on Build GRCh38
rs201322877 Chr.5: 69566599-69566599 on Build GRCh38
rs760411100 Chr.5: 69566544-69566544 on Build GRCh38
rs569458926 Chr.5: 69566603-69566603 on Build GRCh38
rs890229663 Chr.5: 69566355-69566355 on Build GRCh38
rs879122649 Chr.5: 69566537-69566537 on Build GRCh38
rs777108494 Chr.5: 69566550-69566550 on Build GRCh38
rs200500586 Chr.5: 69566536-69566536 on Build GRCh38
rs878879201 Chr.5: 69566388-69566389 on Build GRCh38
rs751760043 Chr.5: 69566614-69566614 on Build GRCh38
rs749047091 Chr.5: 69566649-69566649 on Build GRCh38
rs528456573 Chr.5: 69566460-69566460 on Build GRCh38
rs754745195 Chr.5: 69566668-69566668 on Build GRCh38
rs878899069 Chr.5: 69566543-69566543 on Build GRCh38
rs199618050 Chr.5: 69566525-69566526 on Build GRCh38
rs371199291 Chr.5: 69566535-69566535 on Build GRCh38
rs879249133 Chr.5: 69566537-69566538 on Build GRCh38
rs547690207 Chr.5: 69566503-69566503 on Build GRCh38
rs759954028 Chr.5: 69566555-69566555 on Build GRCh38
rs779711397 Chr.5: 69566618-69566618 on Build GRCh38
rs766626408 Chr.5: 69566545-69566545 on Build GRCh38
rs59898528 Chr.5: 69566538-69566538 on Build GRCh38
rs58303754 Chr.5: 69566535-69566535 on Build GRCh38
rs753215965 Chr.5: 69566578-69566578 on Build GRCh38
rs2561157 Chr.5: 69566533-69566534 on Build GRCh38
rs335259 Chr.5: 69566476-69566476 on Build GRCh38
rs543092995 Chr.5: 69566377-69566377 on Build GRCh38
rs1047459256 Chr.5: 69566323-69566323 on Build GRCh38
rs765663651 Chr.5: 69566562-69566562 on Build GRCh38

View Full Product Details