Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74706286-74706636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2I
Entrez Gene ID
2969
Gene Name
general transcription factor IIi
Gene Aliases
BAP135, BTKAP1, DIWS, GTFII-I, IB291, SPIN, TFII-I, WBS, WBSCR6
UniGene
Hs.647041
Gene Chromosome Location
Chr.7: 74657665-74760692 on Build GRCh38
Transcripts
NM_001163636.2 NM_001280800.1 NM_001518.4 NM_032999.3 NM_033000.3 NM_033001.3 NR_110044.1

Gene Symbol
LOC101926943
Entrez Gene ID
101926943
Gene Name
uncharacterized LOC101926943
Gene Aliases
-
UniGene
Hs.654772
Gene Chromosome Location
Chr.7: 74688937-74728918 on Build GRCh38
Transcripts
NM_001163636.2 NM_001280800.1 NM_001518.4 NM_032999.3 NM_033000.3 NM_033001.3 NR_110044.1

SNP Detail

SNP ID Chromosome Location
rs781890601 Chr.7: 74706423-74706423 on Build GRCh38
rs782799643 Chr.7: 74706356-74706356 on Build GRCh38
rs957563876 Chr.7: 74706529-74706529 on Build GRCh38
rs372072317 Chr.7: 74706492-74706492 on Build GRCh38
rs782441513 Chr.7: 74706476-74706476 on Build GRCh38
rs782564844 Chr.7: 74706430-74706430 on Build GRCh38
rs782466588 Chr.7: 74706447-74706447 on Build GRCh38
rs911586483 Chr.7: 74706608-74706608 on Build GRCh38
rs944649155 Chr.7: 74706309-74706309 on Build GRCh38
rs782235035 Chr.7: 74706445-74706445 on Build GRCh38
rs966327494 Chr.7: 74706593-74706593 on Build GRCh38
rs782802810 Chr.7: 74706417-74706417 on Build GRCh38
rs886402635 Chr.7: 74706459-74706459 on Build GRCh38
rs587608701 Chr.7: 74706507-74706507 on Build GRCh38
rs782450272 Chr.7: 74706428-74706428 on Build GRCh38
rs112395602 Chr.7: 74706554-74706554 on Build GRCh38
rs781950326 Chr.7: 74706485-74706485 on Build GRCh38
rs782661197 Chr.7: 74706461-74706461 on Build GRCh38
rs782175056 Chr.7: 74706352-74706352 on Build GRCh38
rs781863138 Chr.7: 74706387-74706387 on Build GRCh38
rs141631068 Chr.7: 74706317-74706317 on Build GRCh38
rs1049146200 Chr.7: 74706397-74706397 on Build GRCh38
rs782710163 Chr.7: 74706561-74706561 on Build GRCh38
rs900367378 Chr.7: 74706336-74706336 on Build GRCh38
rs782739189 Chr.7: 74706390-74706390 on Build GRCh38
rs782317768 Chr.7: 74706483-74706483 on Build GRCh38
rs587653629 Chr.7: 74706376-74706376 on Build GRCh38
rs1038830014 Chr.7: 74706310-74706310 on Build GRCh38
rs1034930517 Chr.7: 74706513-74706513 on Build GRCh38
rs781986410 Chr.7: 74706345-74706345 on Build GRCh38
rs371779138 Chr.7: 74706460-74706460 on Build GRCh38
rs782240511 Chr.7: 74706474-74706474 on Build GRCh38
rs781942868 Chr.7: 74706357-74706357 on Build GRCh38
rs907442861 Chr.7: 74706494-74706494 on Build GRCh38
rs782096649 Chr.7: 74706370-74706370 on Build GRCh38
rs1022059383 Chr.7: 74706577-74706577 on Build GRCh38
rs781829464 Chr.7: 74706600-74706600 on Build GRCh38
rs587697872 Chr.7: 74706609-74706609 on Build GRCh38
rs782341141 Chr.7: 74706369-74706369 on Build GRCh38

View Full Product Details