Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113828518-113828809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS6
Entrez Gene ID
2621
Gene Name
growth arrest specific 6
Gene Aliases
AXLLG, AXSF
UniGene
Hs.646346
Gene Chromosome Location
Chr.13: 113820549-113864103 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Gene Symbol
GAS6-AS1
Entrez Gene ID
650669
Gene Name
GAS6 antisense RNA 1
Gene Aliases
-
UniGene
Hs.369201
Gene Chromosome Location
Chr.13: 113815610-113842841 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470129 c.1126G>A p.E376K Chr.13: 113828567-113828567 on Build GRCh38
1651268 c.1390C>A p.L464M Chr.13: 113828594-113828594 on Build GRCh38
6669447 c.1074G>A p.P358P Chr.13: 113828619-113828619 on Build GRCh38
4777028 c.1282G>T p.E428* Chr.13: 113828702-113828702 on Build GRCh38
6351877 c.1263G>C p.L421L Chr.13: 113828592-113828592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760562323 Chr.13: 113828698-113828698 on Build GRCh38
rs984375367 Chr.13: 113828625-113828625 on Build GRCh38
rs756300314 Chr.13: 113828646-113828646 on Build GRCh38
rs143062446 Chr.13: 113828637-113828637 on Build GRCh38
rs779439661 Chr.13: 113828617-113828617 on Build GRCh38
rs767448147 Chr.13: 113828645-113828645 on Build GRCh38
rs758697386 Chr.13: 113828664-113828664 on Build GRCh38
rs371632616 Chr.13: 113828588-113828588 on Build GRCh38
rs149411733 Chr.13: 113828600-113828600 on Build GRCh38
rs374326361 Chr.13: 113828549-113828549 on Build GRCh38
rs761345030 Chr.13: 113828556-113828556 on Build GRCh38
rs769290640 Chr.13: 113828636-113828636 on Build GRCh38
rs959989670 Chr.13: 113828785-113828785 on Build GRCh38
rs748784454 Chr.13: 113828689-113828689 on Build GRCh38
rs144103562 Chr.13: 113828661-113828661 on Build GRCh38
rs74118432 Chr.13: 113828751-113828751 on Build GRCh38
rs774477272 Chr.13: 113828643-113828643 on Build GRCh38
rs8191975 Chr.13: 113828592-113828592 on Build GRCh38
rs773143110 Chr.13: 113828576-113828576 on Build GRCh38
rs113292330 Chr.13: 113828614-113828614 on Build GRCh38
rs775130717 Chr.13: 113828697-113828697 on Build GRCh38
rs777715890 Chr.13: 113828605-113828605 on Build GRCh38
rs776227147 Chr.13: 113828622-113828622 on Build GRCh38
rs769326028 Chr.13: 113828675-113828675 on Build GRCh38
rs762589571 Chr.13: 113828638-113828638 on Build GRCh38
rs202080371 Chr.13: 113828555-113828555 on Build GRCh38
rs751838791 Chr.13: 113828749-113828749 on Build GRCh38
rs139784202 Chr.13: 113828619-113828619 on Build GRCh38
rs1036127726 Chr.13: 113828741-113828741 on Build GRCh38
rs1036111444 Chr.13: 113828562-113828562 on Build GRCh38
rs764203956 Chr.13: 113828715-113828715 on Build GRCh38
rs746188272 Chr.13: 113828574-113828578 on Build GRCh38
rs754114986 Chr.13: 113828726-113828726 on Build GRCh38
rs772450271 Chr.13: 113828620-113828620 on Build GRCh38
rs370186377 Chr.13: 113828746-113828746 on Build GRCh38
rs766387009 Chr.13: 113828639-113828639 on Build GRCh38
rs747761635 Chr.13: 113828635-113828635 on Build GRCh38
rs759948442 Chr.13: 113828538-113828538 on Build GRCh38
rs747464759 Chr.13: 113828674-113828674 on Build GRCh38
rs780296101 Chr.13: 113828673-113828673 on Build GRCh38
rs142494562 Chr.13: 113828687-113828687 on Build GRCh38
rs770585835 Chr.13: 113828690-113828690 on Build GRCh38
rs753834414 Chr.13: 113828649-113828649 on Build GRCh38
rs771931132 Chr.13: 113828692-113828692 on Build GRCh38
rs767823966 Chr.13: 113828598-113828598 on Build GRCh38
rs371923342 Chr.13: 113828663-113828663 on Build GRCh38
rs765307266 Chr.13: 113828727-113828727 on Build GRCh38
rs375788560 Chr.13: 113828691-113828691 on Build GRCh38
rs368960262 Chr.13: 113828602-113828602 on Build GRCh38
rs575809095 Chr.13: 113828561-113828561 on Build GRCh38
rs754859745 Chr.13: 113828595-113828595 on Build GRCh38
rs750872309 Chr.13: 113828730-113828730 on Build GRCh38
rs933661454 Chr.13: 113828655-113828655 on Build GRCh38
rs758893786 Chr.13: 113828731-113828731 on Build GRCh38
rs777240223 Chr.13: 113828752-113828752 on Build GRCh38
rs146140804 Chr.13: 113828644-113828644 on Build GRCh38
rs374659031 Chr.13: 113828589-113828589 on Build GRCh38
rs749740507 Chr.13: 113828606-113828606 on Build GRCh38
rs944315569 Chr.13: 113828714-113828714 on Build GRCh38

View Full Product Details