Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18070372-18070662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12RB1
Entrez Gene ID
3594
Gene Name
interleukin 12 receptor subunit beta 1
Gene Aliases
CD212, IL-12R-BETA1, IL12RB, IMD30
UniGene
Hs.567294
Gene Chromosome Location
Chr.19: 18058994-18099027 on Build GRCh38
Transcripts
NM_001290023.1 NM_001290024.1 NM_005535.2 NM_153701.2 XM_006722741.3 XM_011527966.2 XM_011527967.2 XM_011527968.2 XM_011527969.2 XM_011527970.2 XM_011527971.2 XM_011527972.2 XM_011527973.2 XM_011527974.2 XM_011527975.2 XM_011527976.2 XM_011527977.2 XM_017026762.1

SNP Detail

SNP ID Chromosome Location
rs964442371 Chr.19: 18070427-18070427 on Build GRCh38
rs184671668 Chr.19: 18070600-18070600 on Build GRCh38
rs1047401421 Chr.19: 18070459-18070459 on Build GRCh38
rs997600379 Chr.19: 18070638-18070638 on Build GRCh38
rs365179 Chr.19: 18070468-18070468 on Build GRCh38
rs528362889 Chr.19: 18070578-18070578 on Build GRCh38
rs937953025 Chr.19: 18070488-18070488 on Build GRCh38
rs1013725051 Chr.19: 18070552-18070552 on Build GRCh38
rs895347240 Chr.19: 18070521-18070521 on Build GRCh38
rs867661941 Chr.19: 18070533-18070533 on Build GRCh38
rs374240192 Chr.19: 18070504-18070504 on Build GRCh38
rs902613394 Chr.19: 18070598-18070598 on Build GRCh38
rs923036237 Chr.19: 18070431-18070431 on Build GRCh38
rs897874831 Chr.19: 18070621-18070621 on Build GRCh38
rs1046436175 Chr.19: 18070584-18070584 on Build GRCh38
rs752246143 Chr.19: 18070601-18070601 on Build GRCh38
rs1006199680 Chr.19: 18070515-18070515 on Build GRCh38
rs1055694382 Chr.19: 18070513-18070513 on Build GRCh38
rs1048287025 Chr.19: 18070507-18070507 on Build GRCh38
rs928929307 Chr.19: 18070443-18070443 on Build GRCh38
rs533176064 Chr.19: 18070453-18070453 on Build GRCh38
rs942337939 Chr.19: 18070503-18070503 on Build GRCh38
rs564206331 Chr.19: 18070428-18070428 on Build GRCh38

View Full Product Details