Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 14866478-14866935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOMO1
Entrez Gene ID
23420
Gene Name
NODAL modulator 1
Gene Aliases
Nomo, PM5
UniGene
Hs.743963
Gene Chromosome Location
Chr.16: 14833786-14896157 on Build GRCh38
Transcripts
NM_014287.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224500 c.1738A>G p.M580V Chr.16: 14866623-14866623 on Build GRCh38
701919 c.1728G>T p.L576L Chr.16: 14866613-14866613 on Build GRCh38
6143459 c.1729G>T p.E577* Chr.16: 14866614-14866614 on Build GRCh38
7105411 c.1778G>C p.R593T Chr.16: 14866663-14866663 on Build GRCh38
6727762 c.1796C>G p.A599G Chr.16: 14866681-14866681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199820098 Chr.16: 14866732-14866732 on Build GRCh38
rs748795277 Chr.16: 14866536-14866536 on Build GRCh38
rs771403265 Chr.16: 14866499-14866499 on Build GRCh38
rs760111217 Chr.16: 14866551-14866551 on Build GRCh38
rs558882834 Chr.16: 14866790-14866790 on Build GRCh38
rs1044700876 Chr.16: 14866738-14866738 on Build GRCh38
rs537389040 Chr.16: 14866749-14866749 on Build GRCh38
rs753737046 Chr.16: 14866719-14866719 on Build GRCh38
rs371692895 Chr.16: 14866520-14866520 on Build GRCh38
rs760963208 Chr.16: 14866515-14866515 on Build GRCh38
rs745415475 Chr.16: 14866656-14866656 on Build GRCh38
rs537610282 Chr.16: 14866779-14866779 on Build GRCh38
rs772759539 Chr.16: 14866562-14866562 on Build GRCh38
rs370006563 Chr.16: 14866733-14866733 on Build GRCh38
rs17356851 Chr.16: 14866623-14866623 on Build GRCh38
rs376682700 Chr.16: 14866532-14866532 on Build GRCh38
rs755880662 Chr.16: 14866629-14866629 on Build GRCh38
rs751510541 Chr.16: 14866521-14866521 on Build GRCh38
rs571474187 Chr.16: 14866519-14866519 on Build GRCh38
rs1005031878 Chr.16: 14866815-14866815 on Build GRCh38
rs753964253 Chr.16: 14866643-14866643 on Build GRCh38
rs189524829 Chr.16: 14866797-14866797 on Build GRCh38
rs781145556 Chr.16: 14866605-14866605 on Build GRCh38
rs542376298 Chr.16: 14866829-14866829 on Build GRCh38
rs768907111 Chr.16: 14866680-14866680 on Build GRCh38
rs571681940 Chr.16: 14866817-14866817 on Build GRCh38
rs765246439 Chr.16: 14866594-14866594 on Build GRCh38
rs201451365 Chr.16: 14866729-14866729 on Build GRCh38
rs773243782 Chr.16: 14866699-14866699 on Build GRCh38
rs987600127 Chr.16: 14866867-14866867 on Build GRCh38
rs200857849 Chr.16: 14866710-14866710 on Build GRCh38
rs770374322 Chr.16: 14866539-14866539 on Build GRCh38
rs921996951 Chr.16: 14866862-14866862 on Build GRCh38
rs767390418 Chr.16: 14866700-14866700 on Build GRCh38
rs369436605 Chr.16: 14866714-14866714 on Build GRCh38
rs375051004 Chr.16: 14866525-14866525 on Build GRCh38
rs538930977 Chr.16: 14866531-14866531 on Build GRCh38
rs765510122 Chr.16: 14866695-14866695 on Build GRCh38
rs774945402 Chr.16: 14866541-14866541 on Build GRCh38
rs764756957 Chr.16: 14866500-14866500 on Build GRCh38
rs764336223 Chr.16: 14866573-14866573 on Build GRCh38
rs566062971 Chr.16: 14866631-14866631 on Build GRCh38
rs754039811 Chr.16: 14866577-14866577 on Build GRCh38
rs376653019 Chr.16: 14866728-14866728 on Build GRCh38
rs780961455 Chr.16: 14866523-14866523 on Build GRCh38
rs577159274 Chr.16: 14866802-14866802 on Build GRCh38
rs935183788 Chr.16: 14866891-14866891 on Build GRCh38
rs541310986 Chr.16: 14866810-14866810 on Build GRCh38
rs146403442 Chr.16: 14866707-14866707 on Build GRCh38
rs954864644 Chr.16: 14866864-14866864 on Build GRCh38
rs963592442 Chr.16: 14866835-14866835 on Build GRCh38
rs776855089 Chr.16: 14866681-14866681 on Build GRCh38
rs753175664 Chr.16: 14866512-14866512 on Build GRCh38
rs761928018 Chr.16: 14866686-14866686 on Build GRCh38
rs559745568 Chr.16: 14866811-14866811 on Build GRCh38
rs776165910 Chr.16: 14866679-14866679 on Build GRCh38
rs975152668 Chr.16: 14866849-14866849 on Build GRCh38
rs536771392 Chr.16: 14866649-14866649 on Build GRCh38
rs906275902 Chr.16: 14866748-14866748 on Build GRCh38
rs761886720 Chr.16: 14866579-14866579 on Build GRCh38
rs922020177 Chr.16: 14866692-14866692 on Build GRCh38
rs772687680 Chr.16: 14866662-14866662 on Build GRCh38
rs140022331 Chr.16: 14866910-14866910 on Build GRCh38
rs17855981 Chr.16: 14866633-14866633 on Build GRCh38
rs377053115 Chr.16: 14866713-14866713 on Build GRCh38
rs915166044 Chr.16: 14866906-14866906 on Build GRCh38
rs372200759 Chr.16: 14866648-14866648 on Build GRCh38
rs375386997 Chr.16: 14866506-14866506 on Build GRCh38
rs760983671 Chr.16: 14866571-14866571 on Build GRCh38
rs775971944 Chr.16: 14866568-14866568 on Build GRCh38
rs764269574 Chr.16: 14866516-14866516 on Build GRCh38
rs367915791 Chr.16: 14866601-14866601 on Build GRCh38
rs551953081 Chr.16: 14866642-14866642 on Build GRCh38
rs768012370 Chr.16: 14866507-14866507 on Build GRCh38
rs754904035 Chr.16: 14866599-14866599 on Build GRCh38
rs924007481 Chr.16: 14866882-14866882 on Build GRCh38
rs561000273 Chr.16: 14866830-14866830 on Build GRCh38
rs1025387799 Chr.16: 14866755-14866755 on Build GRCh38
rs750559800 Chr.16: 14866596-14866596 on Build GRCh38
rs878915652 Chr.16: 14866827-14866828 on Build GRCh38

View Full Product Details