Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69648077-69648211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND1
Entrez Gene ID
595
Gene Name
cyclin D1
Gene Aliases
BCL1, D11S287E, PRAD1, U21B31
UniGene
Hs.523852
Gene Chromosome Location
Chr.11: 69641105-69654474 on Build GRCh38
Transcripts
NM_053056.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6569030 c.699C>T p.L233L Chr.11: 69648118-69648118 on Build GRCh38
6622956 c.675_677delCTA p.Y227delY Chr.11: 69648094-69648096 on Build GRCh38
1200150 c.697C>T p.L233F Chr.11: 69648116-69648116 on Build GRCh38
1676257 c.718G>C p.D240H Chr.11: 69648137-69648137 on Build GRCh38
4146233 c.723G>A p.P241P Chr.11: 69648142-69648142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760907398 Chr.11: 69648133-69648133 on Build GRCh38
rs745779714 Chr.11: 69648110-69648110 on Build GRCh38
rs773381395 Chr.11: 69648185-69648185 on Build GRCh38
rs9344 Chr.11: 69648142-69648142 on Build GRCh38
rs753117077 Chr.11: 69648187-69648187 on Build GRCh38
rs757978095 Chr.11: 69648189-69648189 on Build GRCh38
rs777225097 Chr.11: 69648141-69648141 on Build GRCh38
rs755236431 Chr.11: 69648155-69648155 on Build GRCh38
rs762019444 Chr.11: 69648112-69648112 on Build GRCh38
rs773470948 Chr.11: 69648130-69648130 on Build GRCh38
rs200179137 Chr.11: 69648102-69648102 on Build GRCh38
rs537247673 Chr.11: 69648176-69648176 on Build GRCh38
rs984643266 Chr.11: 69648108-69648108 on Build GRCh38
rs371455093 Chr.11: 69648152-69648152 on Build GRCh38
rs528354355 Chr.11: 69648175-69648175 on Build GRCh38
rs931750386 Chr.11: 69648158-69648158 on Build GRCh38

View Full Product Details