Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 25089339-25089509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF521
Entrez Gene ID
25925
Gene Name
zinc finger protein 521
Gene Aliases
EHZF, Evi3
UniGene
Hs.116935
Gene Chromosome Location
Chr.18: 25061924-25352250 on Build GRCh38
Transcripts
NM_001308225.1 NM_015461.2 XM_011525909.2 XM_011525911.1 XM_017025697.1 XM_017025698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3524653 c.3874C>T p.P1292S Chr.18: 25089497-25089497 on Build GRCh38
5854016 c.3906+1G>C p.? Chr.18: 25089464-25089464 on Build GRCh38
4800140 c.3898G>T p.E1300* Chr.18: 25089473-25089473 on Build GRCh38
5758792 c.3900G>C p.E1300D Chr.18: 25089471-25089471 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776540373 Chr.18: 25089441-25089442 on Build GRCh38
rs746663273 Chr.18: 25089443-25089443 on Build GRCh38
rs1014312217 Chr.18: 25089407-25089407 on Build GRCh38
rs770559597 Chr.18: 25089449-25089449 on Build GRCh38
rs771934371 Chr.18: 25089422-25089422 on Build GRCh38
rs748454437 Chr.18: 25089379-25089379 on Build GRCh38
rs776338198 Chr.18: 25089461-25089461 on Build GRCh38
rs770782556 Chr.18: 25089417-25089419 on Build GRCh38
rs139036527 Chr.18: 25089363-25089363 on Build GRCh38
rs1023983560 Chr.18: 25089445-25089445 on Build GRCh38
rs748364467 Chr.18: 25089416-25089416 on Build GRCh38
rs760063735 Chr.18: 25089468-25089468 on Build GRCh38
rs772951807 Chr.18: 25089437-25089437 on Build GRCh38
rs997143156 Chr.18: 25089382-25089382 on Build GRCh38

View Full Product Details