Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136497385-136497559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOTCH1
Entrez Gene ID
4851
Gene Name
notch 1
Gene Aliases
AOS5, AOVD1, TAN1, hN1
UniGene
Hs.495473
Gene Chromosome Location
Chr.9: 136494433-136545786 on Build GRCh38
Transcripts
NM_017617.4 XM_011518717.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3215863 c.6295G>A p.D2099N Chr.9: 136497444-136497444 on Build GRCh38
3215865 c.6293G>A p.R2098H Chr.9: 136497446-136497446 on Build GRCh38
6983786 c.6277_6278delAT p.M2093fs*174 Chr.9: 136497461-136497462 on Build GRCh38
3215855 c.6321C>T p.H2107H Chr.9: 136497418-136497418 on Build GRCh38
6914326 c.6310C>T p.R2104C Chr.9: 136497429-136497429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766745955 Chr.9: 136497444-136497444 on Build GRCh38
rs749262096 Chr.9: 136497474-136497474 on Build GRCh38
rs759749364 Chr.9: 136497510-136497510 on Build GRCh38
rs748138179 Chr.9: 136497426-136497426 on Build GRCh38
rs1011153565 Chr.9: 136497490-136497490 on Build GRCh38
rs532201830 Chr.9: 136497418-136497418 on Build GRCh38
rs759998676 Chr.9: 136497446-136497446 on Build GRCh38
rs543842355 Chr.9: 136497463-136497463 on Build GRCh38
rs373091110 Chr.9: 136497425-136497425 on Build GRCh38
rs777022162 Chr.9: 136497508-136497508 on Build GRCh38
rs567909904 Chr.9: 136497534-136497534 on Build GRCh38
rs756964890 Chr.9: 136497412-136497412 on Build GRCh38
rs786205600 Chr.9: 136497539-136497539 on Build GRCh38
rs369665514 Chr.9: 136497423-136497423 on Build GRCh38
rs763651579 Chr.9: 136497451-136497451 on Build GRCh38
rs896859851 Chr.9: 136497430-136497430 on Build GRCh38
rs549258808 Chr.9: 136497516-136497516 on Build GRCh38
rs768543030 Chr.9: 136497479-136497479 on Build GRCh38
rs756874994 Chr.9: 136497455-136497455 on Build GRCh38
rs201058656 Chr.9: 136497482-136497482 on Build GRCh38
rs780963597 Chr.9: 136497417-136497417 on Build GRCh38
rs761295801 Chr.9: 136497526-136497526 on Build GRCh38
rs773483690 Chr.9: 136497439-136497439 on Build GRCh38
rs993857521 Chr.9: 136497434-136497434 on Build GRCh38
rs751048658 Chr.9: 136497530-136497530 on Build GRCh38
rs376501190 Chr.9: 136497436-136497436 on Build GRCh38
rs373806373 Chr.9: 136497480-136497480 on Build GRCh38
rs543533126 Chr.9: 136497441-136497441 on Build GRCh38
rs201987555 Chr.9: 136497448-136497448 on Build GRCh38
rs894965666 Chr.9: 136497411-136497411 on Build GRCh38
rs786205599 Chr.9: 136497486-136497486 on Build GRCh38
rs764551640 Chr.9: 136497517-136497517 on Build GRCh38
rs1022510242 Chr.9: 136497512-136497512 on Build GRCh38
rs531156367 Chr.9: 136497511-136497511 on Build GRCh38
rs777111928 Chr.9: 136497445-136497445 on Build GRCh38
rs780873661 Chr.9: 136497456-136497456 on Build GRCh38
rs765618237 Chr.9: 136497447-136497447 on Build GRCh38
rs747133201 Chr.9: 136497481-136497481 on Build GRCh38
rs767118276 Chr.9: 136497535-136497535 on Build GRCh38
rs755970286 Chr.9: 136497469-136497469 on Build GRCh38

View Full Product Details