Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4276134-4276307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND2
Entrez Gene ID
894
Gene Name
cyclin D2
Gene Aliases
KIAK0002, MPPH3
UniGene
Hs.376071
Gene Chromosome Location
Chr.12: 4273736-4305356 on Build GRCh38
Transcripts
NM_001759.3 NR_125790.1

Gene Symbol
CCND2-AS1
Entrez Gene ID
103752584
Gene Name
CCND2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.577711
Gene Chromosome Location
Chr.12: 4248767-4276184 on Build GRCh38
Transcripts
NM_001759.3 NR_125790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
939326 c.411+1G>T p.? Chr.12: 4276221-4276221 on Build GRCh38
4207380 c.391A>G p.I131V Chr.12: 4276200-4276200 on Build GRCh38
5551557 c.382G>A p.D128N Chr.12: 4276191-4276191 on Build GRCh38
6941170 c.403G>A p.E135K Chr.12: 4276212-4276212 on Build GRCh38
468349 c.329C>G p.S110C Chr.12: 4276138-4276138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767018888 Chr.12: 4276261-4276261 on Build GRCh38
rs867376054 Chr.12: 4276159-4276159 on Build GRCh38
rs774554963 Chr.12: 4276262-4276262 on Build GRCh38
rs769997869 Chr.12: 4276241-4276241 on Build GRCh38
rs777749450 Chr.12: 4276195-4276195 on Build GRCh38
rs766500677 Chr.12: 4276161-4276161 on Build GRCh38
rs781280428 Chr.12: 4276233-4276240 on Build GRCh38
rs751723166 Chr.12: 4276178-4276178 on Build GRCh38
rs200547584 Chr.12: 4276259-4276259 on Build GRCh38
rs745404167 Chr.12: 4276229-4276229 on Build GRCh38
rs763529306 Chr.12: 4276160-4276160 on Build GRCh38
rs928577806 Chr.12: 4276277-4276277 on Build GRCh38
rs1006222339 Chr.12: 4276207-4276207 on Build GRCh38
rs958255950 Chr.12: 4276202-4276202 on Build GRCh38
rs773631850 Chr.12: 4276250-4276250 on Build GRCh38
rs771688425 Chr.12: 4276230-4276230 on Build GRCh38
rs755925937 Chr.12: 4276194-4276194 on Build GRCh38
rs757187520 Chr.12: 4276218-4276218 on Build GRCh38
rs977221684 Chr.12: 4276265-4276265 on Build GRCh38
rs1055859383 Chr.12: 4276172-4276172 on Build GRCh38
rs748346553 Chr.12: 4276248-4276248 on Build GRCh38
rs184590007 Chr.12: 4276190-4276190 on Build GRCh38
rs773652105 Chr.12: 4276157-4276157 on Build GRCh38
rs867321680 Chr.12: 4276196-4276196 on Build GRCh38
rs530949406 Chr.12: 4276282-4276282 on Build GRCh38
rs765770986 Chr.12: 4276156-4276156 on Build GRCh38
rs748533926 Chr.12: 4276238-4276238 on Build GRCh38
rs775162503 Chr.12: 4276231-4276231 on Build GRCh38
rs368616285 Chr.12: 4276215-4276215 on Build GRCh38
rs372384965 Chr.12: 4276227-4276227 on Build GRCh38

View Full Product Details