Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218673670-218673825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK36
Entrez Gene ID
27148
Gene Name
serine/threonine kinase 36
Gene Aliases
FU
UniGene
Hs.471404
Gene Chromosome Location
Chr.2: 218672026-218702717 on Build GRCh38
Transcripts
NM_001243313.1 NM_015690.4 XM_005246464.1 XM_011510959.1 XM_011510960.1 XM_017003804.1 XM_017003805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3391478 c.183C>T p.P61P Chr.2: 218673723-218673723 on Build GRCh38
5708078 c.194A>G p.H65R Chr.2: 218673734-218673734 on Build GRCh38
3380043 c.170G>A p.G57D Chr.2: 218673710-218673710 on Build GRCh38
5457863 c.148C>T p.R50* Chr.2: 218673688-218673688 on Build GRCh38
3380042 c.149G>A p.R50Q Chr.2: 218673689-218673689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs180843126 Chr.2: 218673707-218673707 on Build GRCh38
rs745562452 Chr.2: 218673797-218673797 on Build GRCh38
rs141304903 Chr.2: 218673690-218673690 on Build GRCh38
rs760698830 Chr.2: 218673758-218673758 on Build GRCh38
rs752600883 Chr.2: 218673763-218673763 on Build GRCh38
rs968047009 Chr.2: 218673736-218673736 on Build GRCh38
rs767714293 Chr.2: 218673746-218673746 on Build GRCh38
rs776115212 Chr.2: 218673724-218673724 on Build GRCh38
rs780939590 Chr.2: 218673791-218673791 on Build GRCh38
rs923265567 Chr.2: 218673721-218673721 on Build GRCh38
rs749838134 Chr.2: 218673726-218673726 on Build GRCh38
rs769448814 Chr.2: 218673717-218673717 on Build GRCh38
rs758418181 Chr.2: 218673769-218673769 on Build GRCh38
rs762101900 Chr.2: 218673741-218673741 on Build GRCh38
rs757033557 Chr.2: 218673783-218673783 on Build GRCh38
rs746627715 Chr.2: 218673706-218673706 on Build GRCh38
rs754807843 Chr.2: 218673766-218673767 on Build GRCh38
rs374611897 Chr.2: 218673716-218673716 on Build GRCh38
rs777665185 Chr.2: 218673701-218673701 on Build GRCh38
rs766495036 Chr.2: 218673760-218673760 on Build GRCh38
rs1050678850 Chr.2: 218673764-218673764 on Build GRCh38
rs999615835 Chr.2: 218673770-218673770 on Build GRCh38
rs933354353 Chr.2: 218673732-218673732 on Build GRCh38
rs774840119 Chr.2: 218673737-218673737 on Build GRCh38
rs749749790 Chr.2: 218673802-218673802 on Build GRCh38
rs758106392 Chr.2: 218673700-218673700 on Build GRCh38
rs1022242195 Chr.2: 218673729-218673729 on Build GRCh38
rs755650138 Chr.2: 218673798-218673798 on Build GRCh38
rs763952076 Chr.2: 218673778-218673778 on Build GRCh38
rs751300269 Chr.2: 218673779-218673779 on Build GRCh38
rs768894566 Chr.2: 218673734-218673734 on Build GRCh38
rs773456557 Chr.2: 218673757-218673757 on Build GRCh38
rs752655945 Chr.2: 218673697-218673697 on Build GRCh38

View Full Product Details