Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48685469-48685629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WAS
Entrez Gene ID
7454
Gene Name
Wiskott-Aldrich syndrome
Gene Aliases
IMD2, SCNX, THC, THC1, WASP, WASPA
UniGene
Hs.2157
Gene Chromosome Location
Chr.X: 48683753-48691427 on Build GRCh38
Transcripts
NM_000377.2 XM_011543977.2 XM_017029786.1

SNP Detail

SNP ID Chromosome Location
rs193922414 Chr.X: 48685583-48685583 on Build GRCh38
rs781799471 Chr.X: 48685558-48685558 on Build GRCh38
rs782716215 Chr.X: 48685553-48685553 on Build GRCh38
rs782527416 Chr.X: 48685543-48685543 on Build GRCh38
rs369059472 Chr.X: 48685505-48685505 on Build GRCh38
rs375987005 Chr.X: 48685502-48685502 on Build GRCh38
rs782656368 Chr.X: 48685554-48685554 on Build GRCh38
rs371738193 Chr.X: 48685501-48685501 on Build GRCh38
rs782605575 Chr.X: 48685586-48685586 on Build GRCh38

View Full Product Details