Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52403180-52403344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAP1
Entrez Gene ID
8314
Gene Name
BRCA1 associated protein 1
Gene Aliases
HUCEP-13, UCHL2, hucep-6
UniGene
Hs.106674
Gene Chromosome Location
Chr.3: 52401004-52410105 on Build GRCh38
Transcripts
NM_004656.3 XM_011534149.2 XM_011534150.2 XM_011534151.2 XM_011534152.1 XM_017007303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738879 c.1825A>G p.S609G Chr.3: 52403203-52403203 on Build GRCh38
6909679 c.1807_1829del23 p.V603fs*32 Chr.3: 52403199-52403221 on Build GRCh38
6980158 c.1829G>A p.R610K Chr.3: 52403199-52403199 on Build GRCh38
6922636 c.1840_1851>T p.G614fs*25 Chr.3: 52403177-52403188 on Build GRCh38
5702532 c.1768_1769insG p.Q590fs*53 Chr.3: 52403259-52403260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545682192 Chr.3: 52403294-52403294 on Build GRCh38
rs752063900 Chr.3: 52403207-52403207 on Build GRCh38
rs372576765 Chr.3: 52403231-52403231 on Build GRCh38
rs147775249 Chr.3: 52403279-52403279 on Build GRCh38
rs576624522 Chr.3: 52403256-52403256 on Build GRCh38
rs375793485 Chr.3: 52403276-52403276 on Build GRCh38
rs144172190 Chr.3: 52403247-52403247 on Build GRCh38
rs144993791 Chr.3: 52403223-52403223 on Build GRCh38
rs751730111 Chr.3: 52403237-52403237 on Build GRCh38
rs759423683 Chr.3: 52403222-52403222 on Build GRCh38
rs770654975 Chr.3: 52403208-52403208 on Build GRCh38
rs374877744 Chr.3: 52403277-52403277 on Build GRCh38
rs556046793 Chr.3: 52403238-52403238 on Build GRCh38
rs769728686 Chr.3: 52403308-52403308 on Build GRCh38
rs780338105 Chr.3: 52403265-52403265 on Build GRCh38
rs142059527 Chr.3: 52403227-52403227 on Build GRCh38
rs559555349 Chr.3: 52403311-52403311 on Build GRCh38
rs79014342 Chr.3: 52403242-52403242 on Build GRCh38
rs746575769 Chr.3: 52403292-52403292 on Build GRCh38
rs763927840 Chr.3: 52403235-52403235 on Build GRCh38
rs369065146 Chr.3: 52403218-52403218 on Build GRCh38
rs779252716 Chr.3: 52403287-52403287 on Build GRCh38
rs772835408 Chr.3: 52403201-52403201 on Build GRCh38
rs756472919 Chr.3: 52403259-52403259 on Build GRCh38
rs369259771 Chr.3: 52403280-52403280 on Build GRCh38
rs370004702 Chr.3: 52403293-52403293 on Build GRCh38

View Full Product Details