Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50185902-50186061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL1A1
Entrez Gene ID
1277
Gene Name
collagen type I alpha 1
Gene Aliases
EDSC, OI1, OI2, OI3, OI4
UniGene
Hs.172928
Gene Chromosome Location
Chr.17: 50184096-50201648 on Build GRCh38
Transcripts
NM_000088.3 XM_005257058.3 XM_005257059.4 XM_011524341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777567 c.4031A>C p.D1344A Chr.17: 50185995-50185995 on Build GRCh38
4621627 c.4028C>T p.S1343F Chr.17: 50185998-50185998 on Build GRCh38
1750134 c.4069C>T p.L1357L Chr.17: 50185957-50185957 on Build GRCh38
6632736 c.4037C>T p.A1346V Chr.17: 50185989-50185989 on Build GRCh38
2698908 c.4067G>A p.R1356H Chr.17: 50185959-50185959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201256042 Chr.17: 50186009-50186009 on Build GRCh38
rs149820303 Chr.17: 50185959-50185959 on Build GRCh38
rs772033648 Chr.17: 50185986-50185986 on Build GRCh38
rs776648996 Chr.17: 50185995-50185995 on Build GRCh38
rs191166865 Chr.17: 50185987-50185987 on Build GRCh38
rs371547661 Chr.17: 50185996-50185996 on Build GRCh38
rs753417343 Chr.17: 50186028-50186028 on Build GRCh38
rs752203049 Chr.17: 50185942-50185942 on Build GRCh38
rs753536372 Chr.17: 50185946-50185946 on Build GRCh38
rs886053160 Chr.17: 50185934-50185934 on Build GRCh38
rs760040029 Chr.17: 50186017-50186017 on Build GRCh38
rs375650769 Chr.17: 50186031-50186031 on Build GRCh38
rs763676384 Chr.17: 50186018-50186018 on Build GRCh38
rs982770651 Chr.17: 50185968-50185968 on Build GRCh38
rs778734618 Chr.17: 50185984-50185984 on Build GRCh38
rs147936946 Chr.17: 50186008-50186008 on Build GRCh38
rs750267930 Chr.17: 50186038-50186038 on Build GRCh38
rs773753538 Chr.17: 50186002-50186002 on Build GRCh38
rs745797859 Chr.17: 50185985-50185985 on Build GRCh38
rs545518162 Chr.17: 50185965-50185965 on Build GRCh38
rs768810493 Chr.17: 50185988-50185988 on Build GRCh38
rs72656347 Chr.17: 50185972-50185972 on Build GRCh38
rs141011435 Chr.17: 50185945-50185945 on Build GRCh38
rs113005305 Chr.17: 50185962-50185962 on Build GRCh38
rs778644341 Chr.17: 50186032-50186032 on Build GRCh38
rs377703930 Chr.17: 50185964-50185964 on Build GRCh38
rs72656346 Chr.17: 50186013-50186018 on Build GRCh38
rs758211486 Chr.17: 50186040-50186040 on Build GRCh38
rs202122731 Chr.17: 50186036-50186036 on Build GRCh38
rs193922156 Chr.17: 50186029-50186029 on Build GRCh38
rs770279565 Chr.17: 50185997-50185997 on Build GRCh38

View Full Product Details