Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80263553-80263721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF213
Entrez Gene ID
57674
Gene Name
ring finger protein 213
Gene Aliases
ALO17, C17orf27, KIAA1618, MYMY2, MYSTR, NET57
UniGene
Hs.195642
Gene Chromosome Location
Chr.17: 80260820-80398786 on Build GRCh38
Transcripts
NM_001256071.2 NM_020954.3 XM_005257545.4 XM_005257546.4 XM_006721995.3 XM_011525084.2 XM_011525086.2 XM_011525087.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5820128 c.18C>G p.C6W Chr.17: 80263699-80263699 on Build GRCh38
1303468 c.14C>T p.S5L Chr.17: 80263695-80263695 on Build GRCh38
1303467 c.14C>T p.S5L Chr.17: 80263695-80263695 on Build GRCh38
1303469 c.14C>T p.S5L Chr.17: 80263695-80263695 on Build GRCh38
1223965 c.25G>T p.V9F Chr.17: 80263706-80263706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048371583 Chr.17: 80263616-80263616 on Build GRCh38
rs777734027 Chr.17: 80263660-80263660 on Build GRCh38
rs369296898 Chr.17: 80263638-80263638 on Build GRCh38
rs1053265658 Chr.17: 80263615-80263615 on Build GRCh38
rs566320605 Chr.17: 80263679-80263679 on Build GRCh38
rs142962817 Chr.17: 80263695-80263695 on Build GRCh38
rs369918247 Chr.17: 80263644-80263644 on Build GRCh38
rs780595875 Chr.17: 80263633-80263633 on Build GRCh38
rs748240232 Chr.17: 80263645-80263645 on Build GRCh38
rs999438692 Chr.17: 80263605-80263605 on Build GRCh38
rs372376812 Chr.17: 80263696-80263696 on Build GRCh38
rs887139483 Chr.17: 80263618-80263618 on Build GRCh38
rs926709224 Chr.17: 80263590-80263590 on Build GRCh38
rs931434111 Chr.17: 80263608-80263608 on Build GRCh38
rs769753503 Chr.17: 80263657-80263657 on Build GRCh38
rs747177989 Chr.17: 80263634-80263634 on Build GRCh38
rs904736220 Chr.17: 80263591-80263591 on Build GRCh38

View Full Product Details