Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17831299-17831458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
UniGene
Hs.515247
Gene Chromosome Location
Chr.19: 17824782-17848071 on Build GRCh38
Transcripts
NM_000215.3 XM_005259896.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363729 c.2828G>A p.R943H Chr.19: 17831378-17831378 on Build GRCh38
4693748 c.2845G>A p.D949N Chr.19: 17831361-17831361 on Build GRCh38
6932428 c.2878G>A p.E960K Chr.19: 17831328-17831328 on Build GRCh38
6689329 c.2859A>G p.R953R Chr.19: 17831347-17831347 on Build GRCh38
6689317 c.2836G>A p.V946M Chr.19: 17831370-17831370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200202992 Chr.19: 17831364-17831364 on Build GRCh38
rs193156379 Chr.19: 17831413-17831413 on Build GRCh38
rs749741343 Chr.19: 17831334-17831334 on Build GRCh38
rs762179562 Chr.19: 17831352-17831352 on Build GRCh38
rs779843346 Chr.19: 17831320-17831320 on Build GRCh38
rs372733285 Chr.19: 17831381-17831381 on Build GRCh38
rs567601710 Chr.19: 17831341-17831341 on Build GRCh38
rs976222264 Chr.19: 17831414-17831414 on Build GRCh38
rs951360037 Chr.19: 17831361-17831361 on Build GRCh38
rs199553891 Chr.19: 17831344-17831344 on Build GRCh38
rs199653549 Chr.19: 17831420-17831420 on Build GRCh38
rs769113856 Chr.19: 17831338-17831338 on Build GRCh38
rs766654268 Chr.19: 17831403-17831403 on Build GRCh38
rs760901179 Chr.19: 17831379-17831379 on Build GRCh38

View Full Product Details