Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125626688-125626859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHEK1
Entrez Gene ID
1111
Gene Name
checkpoint kinase 1
Gene Aliases
CHK1
UniGene
Hs.24529
Gene Chromosome Location
Chr.11: 125624910-125676259 on Build GRCh38
Transcripts
NM_001114121.2 NM_001114122.2 NM_001244846.1 NM_001274.5 NR_045204.1 NR_045205.1 XM_011542560.2 XM_011542561.2 XM_011542562.2 XM_011542563.2 XM_017017146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5104695 c.65+1G>A p.? Chr.11: 125626834-125626834 on Build GRCh38
4761052 c.41C>A p.T14N Chr.11: 125626809-125626809 on Build GRCh38
5717631 c.12C>T p.P4P Chr.11: 125626780-125626780 on Build GRCh38
6960065 c.5C>T p.A2V Chr.11: 125626773-125626773 on Build GRCh38
4990139 c.19G>A p.E7K Chr.11: 125626787-125626787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778209583 Chr.11: 125626715-125626716 on Build GRCh38
rs200520758 Chr.11: 125626802-125626802 on Build GRCh38
rs756973382 Chr.11: 125626764-125626764 on Build GRCh38
rs769608055 Chr.11: 125626796-125626797 on Build GRCh38
rs945101438 Chr.11: 125626710-125626710 on Build GRCh38
rs769358548 Chr.11: 125626720-125626720 on Build GRCh38
rs757321404 Chr.11: 125626773-125626773 on Build GRCh38
rs762669601 Chr.11: 125626729-125626729 on Build GRCh38
rs1056080774 Chr.11: 125626749-125626749 on Build GRCh38
rs556571980 Chr.11: 125626750-125626750 on Build GRCh38
rs774282933 Chr.11: 125626744-125626744 on Build GRCh38
rs766129595 Chr.11: 125626737-125626737 on Build GRCh38
rs780466731 Chr.11: 125626819-125626819 on Build GRCh38
rs772872053 Chr.11: 125626721-125626721 on Build GRCh38
rs926560464 Chr.11: 125626714-125626714 on Build GRCh38
rs754011771 Chr.11: 125626767-125626767 on Build GRCh38
rs144523921 Chr.11: 125626807-125626807 on Build GRCh38
rs189276906 Chr.11: 125626713-125626713 on Build GRCh38
rs759605889 Chr.11: 125626748-125626748 on Build GRCh38
rs371037025 Chr.11: 125626751-125626751 on Build GRCh38
rs751896993 Chr.11: 125626829-125626829 on Build GRCh38
rs201459198 Chr.11: 125626778-125626778 on Build GRCh38

View Full Product Details