Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 18631557-18631707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAX7
Entrez Gene ID
5081
Gene Name
paired box 7
Gene Aliases
HUP1, PAX7B, RMS2
UniGene
Hs.113253
Gene Chromosome Location
Chr.1: 18631006-18748866 on Build GRCh38
Transcripts
NM_001135254.1 NM_002584.2 NM_013945.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5953306 c.47C>T p.P16L Chr.1: 18631650-18631650 on Build GRCh38
6738264 c.27G>A p.P9P Chr.1: 18631630-18631630 on Build GRCh38
4560104 c.82G>A p.E28K Chr.1: 18631685-18631685 on Build GRCh38
5953305 c.47C>T p.P16L Chr.1: 18631650-18631650 on Build GRCh38
6738279 c.60delC p.R22fs*38 Chr.1: 18631663-18631663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548308018 Chr.1: 18631642-18631642 on Build GRCh38
rs1033982888 Chr.1: 18631650-18631650 on Build GRCh38
rs774720166 Chr.1: 18631648-18631648 on Build GRCh38
rs552417333 Chr.1: 18631668-18631668 on Build GRCh38
rs570631887 Chr.1: 18631679-18631679 on Build GRCh38
rs748250427 Chr.1: 18631629-18631629 on Build GRCh38
rs1053554548 Chr.1: 18631628-18631628 on Build GRCh38
rs562546072 Chr.1: 18631609-18631609 on Build GRCh38
rs776238209 Chr.1: 18631659-18631659 on Build GRCh38
rs746324287 Chr.1: 18631649-18631649 on Build GRCh38
rs566629103 Chr.1: 18631644-18631644 on Build GRCh38
rs754895481 Chr.1: 18631682-18631682 on Build GRCh38
rs771388741 Chr.1: 18631645-18631645 on Build GRCh38
rs774974785 Chr.1: 18631595-18631595 on Build GRCh38
rs191271197 Chr.1: 18631584-18631584 on Build GRCh38
rs763408412 Chr.1: 18631591-18631591 on Build GRCh38
rs761503724 Chr.1: 18631600-18631600 on Build GRCh38
rs751581027 Chr.1: 18631620-18631620 on Build GRCh38
rs900671673 Chr.1: 18631607-18631607 on Build GRCh38
rs1010934600 Chr.1: 18631608-18631608 on Build GRCh38
rs764959341 Chr.1: 18631667-18631667 on Build GRCh38
rs1020589052 Chr.1: 18631663-18631663 on Build GRCh38
rs763731260 Chr.1: 18631597-18631597 on Build GRCh38
rs200862549 Chr.1: 18631660-18631660 on Build GRCh38
rs762651087 Chr.1: 18631671-18631671 on Build GRCh38
rs1020540728 Chr.1: 18631614-18631614 on Build GRCh38
rs771314639 Chr.1: 18631593-18631593 on Build GRCh38
rs770017663 Chr.1: 18631577-18631577 on Build GRCh38
rs758262026 Chr.1: 18631618-18631618 on Build GRCh38
rs753408357 Chr.1: 18631598-18631598 on Build GRCh38
rs377198222 Chr.1: 18631654-18631654 on Build GRCh38
rs778012759 Chr.1: 18631643-18631643 on Build GRCh38
rs144835222 Chr.1: 18631624-18631624 on Build GRCh38
rs529809505 Chr.1: 18631611-18631611 on Build GRCh38

View Full Product Details