Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 89946106-89946279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBN
Entrez Gene ID
4683
Gene Name
nibrin
Gene Aliases
AT-V1, AT-V2, ATV, NBS, NBS1, P95
UniGene
Hs.492208
Gene Chromosome Location
Chr.8: 89933336-89984733 on Build GRCh38
Transcripts
NM_001024688.2 NM_002485.4 XM_011517045.1 XM_017013460.1 XM_017013461.1 XM_017013462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6959238 c.1939A>G p.S647G Chr.8: 89946271-89946271 on Build GRCh38
6961114 c.2000C>G p.S667C Chr.8: 89946210-89946210 on Build GRCh38
3901967 c.2025A>G p.I675M Chr.8: 89946185-89946185 on Build GRCh38
6845518 c.1969A>G p.T657A Chr.8: 89946241-89946241 on Build GRCh38
1458548 c.1958_1959insA p.L654fs*5 Chr.8: 89946251-89946252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779048193 Chr.8: 89946141-89946141 on Build GRCh38
rs571803328 Chr.8: 89946224-89946224 on Build GRCh38
rs201781110 Chr.8: 89946231-89946231 on Build GRCh38
rs202198205 Chr.8: 89946193-89946193 on Build GRCh38
rs876660950 Chr.8: 89946136-89946136 on Build GRCh38
rs876658336 Chr.8: 89946207-89946207 on Build GRCh38
rs587782653 Chr.8: 89946211-89946224 on Build GRCh38
rs876658655 Chr.8: 89946227-89946227 on Build GRCh38
rs757753217 Chr.8: 89946221-89946221 on Build GRCh38
rs758675398 Chr.8: 89946149-89946149 on Build GRCh38
rs876658488 Chr.8: 89946202-89946202 on Build GRCh38
rs370295427 Chr.8: 89946174-89946174 on Build GRCh38
rs758728938 Chr.8: 89946249-89946249 on Build GRCh38
rs757339440 Chr.8: 89946133-89946133 on Build GRCh38
rs746135009 Chr.8: 89946148-89946148 on Build GRCh38
rs374638426 Chr.8: 89946210-89946210 on Build GRCh38
rs769252040 Chr.8: 89946176-89946176 on Build GRCh38
rs587780092 Chr.8: 89946142-89946142 on Build GRCh38
rs200399787 Chr.8: 89946203-89946203 on Build GRCh38
rs746764974 Chr.8: 89946153-89946153 on Build GRCh38
rs864622555 Chr.8: 89946173-89946173 on Build GRCh38
rs730881856 Chr.8: 89946181-89946181 on Build GRCh38
rs186371605 Chr.8: 89946150-89946150 on Build GRCh38
rs200564603 Chr.8: 89946172-89946172 on Build GRCh38
rs780235686 Chr.8: 89946251-89946252 on Build GRCh38
rs1057516668 Chr.8: 89946236-89946236 on Build GRCh38
rs1057516852 Chr.8: 89946209-89946210 on Build GRCh38
rs587780091 Chr.8: 89946211-89946211 on Build GRCh38
rs765458801 Chr.8: 89946155-89946155 on Build GRCh38
rs878854508 Chr.8: 89946244-89946244 on Build GRCh38
rs768715280 Chr.8: 89946157-89946157 on Build GRCh38
rs1061302 Chr.8: 89946194-89946194 on Build GRCh38
rs1057516869 Chr.8: 89946160-89946161 on Build GRCh38
rs1036111786 Chr.8: 89946172-89946173 on Build GRCh38
rs786203223 Chr.8: 89946138-89946138 on Build GRCh38
rs138913151 Chr.8: 89946208-89946208 on Build GRCh38
rs587782155 Chr.8: 89946237-89946237 on Build GRCh38
rs6413508 Chr.8: 89946195-89946195 on Build GRCh38
rs786203920 Chr.8: 89946154-89946154 on Build GRCh38
rs1057517075 Chr.8: 89946138-89946138 on Build GRCh38

View Full Product Details