Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 70113546-70113720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOA2
Entrez Gene ID
10499
Gene Name
nuclear receptor coactivator 2
Gene Aliases
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75
UniGene
Hs.446678
Gene Chromosome Location
Chr.8: 70109762-70405390 on Build GRCh38
Transcripts
NM_001321703.1 NM_001321707.1 NM_001321711.1 NM_001321712.1 NM_001321713.1 NM_006540.3 XM_017012961.1 XM_017012962.1 XM_017012963.1 XM_017012964.1 XM_017012965.1 XM_017012966.1 XM_017012967.1 XM_017012968.1 XM_017012969.1 XM_017012970.1 XM_017012971.1 XM_017012972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5719080 c.4384-2A>G p.? Chr.8: 70113645-70113645 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901423800 Chr.8: 70113690-70113690 on Build GRCh38
rs774471025 Chr.8: 70113613-70113614 on Build GRCh38
rs528705643 Chr.8: 70113577-70113577 on Build GRCh38
rs76809412 Chr.8: 70113599-70113599 on Build GRCh38
rs750129002 Chr.8: 70113647-70113647 on Build GRCh38
rs772301326 Chr.8: 70113663-70113663 on Build GRCh38
rs759869023 Chr.8: 70113650-70113650 on Build GRCh38
rs978416691 Chr.8: 70113627-70113627 on Build GRCh38
rs947676098 Chr.8: 70113668-70113668 on Build GRCh38
rs746958948 Chr.8: 70113660-70113660 on Build GRCh38
rs751017134 Chr.8: 70113666-70113666 on Build GRCh38
rs765840695 Chr.8: 70113665-70113665 on Build GRCh38
rs368655450 Chr.8: 70113630-70113630 on Build GRCh38
rs913011200 Chr.8: 70113579-70113579 on Build GRCh38
rs774821713 Chr.8: 70113604-70113604 on Build GRCh38
rs910849890 Chr.8: 70113636-70113636 on Build GRCh38
rs765559315 Chr.8: 70113641-70113641 on Build GRCh38

View Full Product Details