Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48342591-48342760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RB1
Entrez Gene ID
5925
Gene Name
RB transcriptional corepressor 1
Gene Aliases
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110
UniGene
Hs.408528
Gene Chromosome Location
Chr.13: 48303747-48481890 on Build GRCh38
Transcripts
NM_000321.2 XM_011535171.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
39409 c.265-1G>A p.? Chr.13: 48342598-48342598 on Build GRCh38
6851675 c.350T>C p.F117S Chr.13: 48342684-48342684 on Build GRCh38
1367152 c.380G>A p.S127N Chr.13: 48342714-48342714 on Build GRCh38
4802882 c.287A>T p.K96M Chr.13: 48342621-48342621 on Build GRCh38
5659096 c.361C>T p.Q121* Chr.13: 48342695-48342695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777410222 Chr.13: 48342697-48342697 on Build GRCh38
rs886050268 Chr.13: 48342633-48342633 on Build GRCh38
rs3092882 Chr.13: 48342731-48342732 on Build GRCh38
rs3092881 Chr.13: 48342726-48342726 on Build GRCh38
rs779647793 Chr.13: 48342629-48342629 on Build GRCh38
rs187110786 Chr.13: 48342724-48342724 on Build GRCh38
rs771961195 Chr.13: 48342679-48342679 on Build GRCh38
rs947950104 Chr.13: 48342709-48342709 on Build GRCh38
rs983885759 Chr.13: 48342693-48342693 on Build GRCh38
rs149800437 Chr.13: 48342701-48342701 on Build GRCh38
rs762953095 Chr.13: 48342720-48342720 on Build GRCh38
rs755688480 Chr.13: 48342619-48342619 on Build GRCh38
rs778932079 Chr.13: 48342670-48342670 on Build GRCh38
rs1052655522 Chr.13: 48342728-48342728 on Build GRCh38
rs951251256 Chr.13: 48342688-48342688 on Build GRCh38
rs587778853 Chr.13: 48342638-48342639 on Build GRCh38
rs376408183 Chr.13: 48342657-48342657 on Build GRCh38
rs139673557 Chr.13: 48342675-48342675 on Build GRCh38
rs767961998 Chr.13: 48342734-48342734 on Build GRCh38
rs753352976 Chr.13: 48342649-48342649 on Build GRCh38
rs869214358 Chr.13: 48342698-48342699 on Build GRCh38
rs775559506 Chr.13: 48342713-48342713 on Build GRCh38
rs769858139 Chr.13: 48342704-48342704 on Build GRCh38
rs576233044 Chr.13: 48342667-48342667 on Build GRCh38
rs774892592 Chr.13: 48342725-48342725 on Build GRCh38
rs587778825 Chr.13: 48342639-48342640 on Build GRCh38
rs748093967 Chr.13: 48342676-48342676 on Build GRCh38
rs794727481 Chr.13: 48342631-48342631 on Build GRCh38
rs919345517 Chr.13: 48342625-48342625 on Build GRCh38
rs886042357 Chr.13: 48342710-48342710 on Build GRCh38

View Full Product Details