Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177230707-177230850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFE2L2
Entrez Gene ID
4780
Gene Name
nuclear factor, erythroid 2 like 2
Gene Aliases
HEBP1, NRF2
UniGene
Hs.744006
Gene Chromosome Location
Chr.2: 177230303-177265131 on Build GRCh38
Transcripts
NM_001145412.3 NM_001145413.3 NM_001313900.1 NM_001313901.1 NM_001313902.1 NM_001313903.1 NM_001313904.1 NM_006164.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5498813 c.1767C>T p.G589G Chr.2: 177230836-177230836 on Build GRCh38
7135079 c.1799C>G p.P600R Chr.2: 177230804-177230804 on Build GRCh38
5671711 c.1791_1794delTAAG p.S597fs*>8 Chr.2: 177230809-177230812 on Build GRCh38
6954934 c.1811A>C p.K604T Chr.2: 177230792-177230792 on Build GRCh38
3837466 c.1800A>G p.P600P Chr.2: 177230803-177230803 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs976522151 Chr.2: 177230817-177230817 on Build GRCh38
rs1011806077 Chr.2: 177230734-177230734 on Build GRCh38
rs1001735936 Chr.2: 177230738-177230738 on Build GRCh38
rs201824058 Chr.2: 177230750-177230750 on Build GRCh38
rs758079733 Chr.2: 177230777-177230777 on Build GRCh38
rs1015105997 Chr.2: 177230792-177230792 on Build GRCh38
rs748937667 Chr.2: 177230809-177230812 on Build GRCh38
rs893301872 Chr.2: 177230733-177230737 on Build GRCh38
rs558313759 Chr.2: 177230794-177230794 on Build GRCh38
rs764648073 Chr.2: 177230740-177230740 on Build GRCh38
rs983989560 Chr.2: 177230773-177230773 on Build GRCh38
rs779911242 Chr.2: 177230786-177230786 on Build GRCh38
rs77547666 Chr.2: 177230767-177230767 on Build GRCh38
rs1049953654 Chr.2: 177230766-177230766 on Build GRCh38
rs753898838 Chr.2: 177230746-177230746 on Build GRCh38
rs761245040 Chr.2: 177230735-177230735 on Build GRCh38
rs765413516 Chr.2: 177230759-177230759 on Build GRCh38
rs769505567 Chr.2: 177230746-177230747 on Build GRCh38
rs754939223 Chr.2: 177230812-177230812 on Build GRCh38
rs777573333 Chr.2: 177230774-177230776 on Build GRCh38

View Full Product Details