Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120978656-120978795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNF1A
Entrez Gene ID
6927
Gene Name
HNF1 homeobox A
Gene Aliases
HNF-1A, HNF1, IDDM20, LFB1, MODY3, TCF-1, TCF1
UniGene
Hs.654455
Gene Chromosome Location
Chr.12: 120978515-121002512 on Build GRCh38
Transcripts
NM_000545.6 NM_001306179.1 XM_005253931.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2175424 c.5T>C p.V2A Chr.12: 120978773-120978773 on Build GRCh38
21479 c.18_36>T p.Q7_L12delQLQTEL Chr.12: 120978786-120978804 on Build GRCh38
7129362 c.15G>C p.L5L Chr.12: 120978783-120978783 on Build GRCh38
21480 c.27_33delGACGGAG p.Q9fs*11 Chr.12: 120978795-120978801 on Build GRCh38
4801483 c.13C>T p.L5L Chr.12: 120978781-120978781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039044797 Chr.12: 120978742-120978765 on Build GRCh38
rs765037128 Chr.12: 120978747-120978747 on Build GRCh38
rs193922592 Chr.12: 120978769-120978769 on Build GRCh38
rs868538299 Chr.12: 120978698-120978698 on Build GRCh38
rs1038779132 Chr.12: 120978703-120978703 on Build GRCh38
rs747774978 Chr.12: 120978704-120978706 on Build GRCh38
rs199945468 Chr.12: 120978746-120978746 on Build GRCh38
rs769332225 Chr.12: 120978710-120978714 on Build GRCh38
rs779387337 Chr.12: 120978763-120978763 on Build GRCh38
rs764010995 Chr.12: 120978745-120978745 on Build GRCh38
rs771466066 Chr.12: 120978712-120978712 on Build GRCh38
rs1033416844 Chr.12: 120978709-120978709 on Build GRCh38
rs778233597 Chr.12: 120978767-120978767 on Build GRCh38
rs752573339 Chr.12: 120978687-120978687 on Build GRCh38
rs748532681 Chr.12: 120978721-120978721 on Build GRCh38
rs1022343054 Chr.12: 120978684-120978684 on Build GRCh38
rs773528592 Chr.12: 120978722-120978722 on Build GRCh38
rs950999882 Chr.12: 120978733-120978733 on Build GRCh38
rs781754738 Chr.12: 120978715-120978715 on Build GRCh38
rs759514603 Chr.12: 120978679-120978679 on Build GRCh38
rs751680667 Chr.12: 120978677-120978677 on Build GRCh38
rs777653568 Chr.12: 120978706-120978706 on Build GRCh38
rs757926841 Chr.12: 120978758-120978758 on Build GRCh38
rs1026580886 Chr.12: 120978726-120978726 on Build GRCh38
rs767550584 Chr.12: 120978680-120978680 on Build GRCh38
rs561269721 Chr.12: 120978765-120978765 on Build GRCh38
rs753567412 Chr.12: 120978707-120978707 on Build GRCh38
rs749712370 Chr.12: 120978768-120978768 on Build GRCh38
rs749624286 Chr.12: 120978724-120978724 on Build GRCh38
rs370979090 Chr.12: 120978764-120978764 on Build GRCh38
rs1047425321 Chr.12: 120978714-120978714 on Build GRCh38
rs759730121 Chr.12: 120978739-120978739 on Build GRCh38
rs755941887 Chr.12: 120978688-120978688 on Build GRCh38
rs545740321 Chr.12: 120978710-120978710 on Build GRCh38
rs774545180 Chr.12: 120978738-120978738 on Build GRCh38
rs565546289 Chr.12: 120978729-120978729 on Build GRCh38
rs376961226 Chr.12: 120978740-120978740 on Build GRCh38
rs778674831 Chr.12: 120978708-120978708 on Build GRCh38
rs750035345 Chr.12: 120978755-120978755 on Build GRCh38
rs775511041 Chr.12: 120978744-120978744 on Build GRCh38

View Full Product Details