Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127271297-127271457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC3
Entrez Gene ID
2071
Gene Name
ERCC excision repair 3, TFIIH core complex helicase subunit
Gene Aliases
BTF2, GTF2H, RAD25, TFIIH, TTD2, XPB
UniGene
Hs.469872
Gene Chromosome Location
Chr.2: 127257290-127294176 on Build GRCh38
Transcripts
NM_000122.1 NM_001303416.1 NM_001303418.1 XM_011510794.1 XM_011510795.1 XM_017003583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1691015 c.1841C>T p.S614L Chr.2: 127271440-127271440 on Build GRCh38
4084668 c.1923G>A p.G641G Chr.2: 127271358-127271358 on Build GRCh38
6909852 c.1936G>C p.A646P Chr.2: 127271345-127271345 on Build GRCh38
5989445 c.1930C>T p.L644F Chr.2: 127271351-127271351 on Build GRCh38
6944735 c.1873C>G p.Q625E Chr.2: 127271408-127271408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053657969 Chr.2: 127271319-127271319 on Build GRCh38
rs765724086 Chr.2: 127271427-127271427 on Build GRCh38
rs184374356 Chr.2: 127271326-127271326 on Build GRCh38
rs758820876 Chr.2: 127271319-127271323 on Build GRCh38
rs775316471 Chr.2: 127271380-127271380 on Build GRCh38
rs748753777 Chr.2: 127271325-127271325 on Build GRCh38
rs767273632 Chr.2: 127271368-127271368 on Build GRCh38
rs760870346 Chr.2: 127271388-127271388 on Build GRCh38
rs865999173 Chr.2: 127271409-127271409 on Build GRCh38
rs780186679 Chr.2: 127271327-127271330 on Build GRCh38
rs753886932 Chr.2: 127271395-127271395 on Build GRCh38
rs757293615 Chr.2: 127271420-127271420 on Build GRCh38
rs774181315 Chr.2: 127271355-127271355 on Build GRCh38
rs958927759 Chr.2: 127271394-127271394 on Build GRCh38
rs759338358 Chr.2: 127271357-127271357 on Build GRCh38
rs764362224 Chr.2: 127271390-127271390 on Build GRCh38
rs369447216 Chr.2: 127271361-127271361 on Build GRCh38
rs375556869 Chr.2: 127271352-127271352 on Build GRCh38

View Full Product Details