Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5501741-5501896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP1
Entrez Gene ID
22861
Gene Name
NLR family pyrin domain containing 1
Gene Aliases
CARD7, CIDED, CLR17.1, DEFCAP, DEFCAP-L/S, NAC, NALP1, PP1044, SLEV1, VAMAS1
UniGene
Hs.652273
Gene Chromosome Location
Chr.17: 5501399-5584512 on Build GRCh38
Transcripts
NM_001033053.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
981589 c.4103G>A p.C1368Y Chr.17: 5501839-5501839 on Build GRCh38
6465009 c.4089G>A p.P1363P Chr.17: 5501853-5501853 on Build GRCh38
1521642 c.4113C>T p.D1371D Chr.17: 5501829-5501829 on Build GRCh38
4384878 c.4117A>G p.R1373G Chr.17: 5501825-5501825 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779427897 Chr.17: 5501791-5501791 on Build GRCh38
rs759297923 Chr.17: 5501848-5501848 on Build GRCh38
rs767309323 Chr.17: 5501850-5501850 on Build GRCh38
rs902546619 Chr.17: 5501767-5501767 on Build GRCh38
rs775032614 Chr.17: 5501852-5501852 on Build GRCh38
rs534096389 Chr.17: 5501781-5501781 on Build GRCh38
rs761407735 Chr.17: 5501866-5501866 on Build GRCh38
rs199661912 Chr.17: 5501824-5501824 on Build GRCh38
rs200674144 Chr.17: 5501835-5501835 on Build GRCh38
rs780193802 Chr.17: 5501814-5501814 on Build GRCh38
rs774257690 Chr.17: 5501837-5501837 on Build GRCh38
rs758780620 Chr.17: 5501810-5501810 on Build GRCh38
rs746305090 Chr.17: 5501806-5501806 on Build GRCh38
rs201762229 Chr.17: 5501788-5501788 on Build GRCh38
rs763615140 Chr.17: 5501769-5501769 on Build GRCh38
rs201026015 Chr.17: 5501831-5501831 on Build GRCh38
rs749226290 Chr.17: 5501829-5501829 on Build GRCh38
rs747224239 Chr.17: 5501817-5501817 on Build GRCh38
rs199812357 Chr.17: 5501864-5501864 on Build GRCh38
rs140863595 Chr.17: 5501854-5501854 on Build GRCh38
rs200802942 Chr.17: 5501828-5501828 on Build GRCh38
rs754416419 Chr.17: 5501780-5501780 on Build GRCh38
rs984668558 Chr.17: 5501853-5501853 on Build GRCh38

View Full Product Details