Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 58709456-58709584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MALT1
Entrez Gene ID
10892
Gene Name
MALT1 paracaspase
Gene Aliases
IMD12, MLT, MLT1, PCASP1
UniGene
Hs.601217
Gene Chromosome Location
Chr.18: 58671386-58753806 on Build GRCh38
Transcripts
NM_006785.3 NM_173844.2 XM_011525794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
330359 c.731T>A p.L244* Chr.18: 58709459-58709459 on Build GRCh38
6954841 c.733G>T p.V245F Chr.18: 58709461-58709461 on Build GRCh38
4420887 c.758G>T p.S253I Chr.18: 58709486-58709486 on Build GRCh38
2880924 c.781T>C p.F261L Chr.18: 58709509-58709509 on Build GRCh38
6926361 c.766C>T p.P256S Chr.18: 58709494-58709494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145169756 Chr.18: 58709552-58709552 on Build GRCh38
rs759593233 Chr.18: 58709532-58709532 on Build GRCh38
rs376106734 Chr.18: 58709526-58709526 on Build GRCh38
rs960844050 Chr.18: 58709554-58709554 on Build GRCh38
rs752497631 Chr.18: 58709541-58709541 on Build GRCh38
rs1033929290 Chr.18: 58709534-58709535 on Build GRCh38
rs762795672 Chr.18: 58709550-58709550 on Build GRCh38
rs765186001 Chr.18: 58709538-58709538 on Build GRCh38

View Full Product Details