Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134172249-134172422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGK1
Entrez Gene ID
6446
Gene Name
serum/glucocorticoid regulated kinase 1
Gene Aliases
SGK
UniGene
Hs.510078
Gene Chromosome Location
Chr.6: 134169246-134318112 on Build GRCh38
Transcripts
NM_001143676.1 NM_001143677.1 NM_001143678.1 NM_001291995.1 NM_005627.3 XM_011536071.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1487238 c.737C>A p.S246* Chr.6: 134172284-134172284 on Build GRCh38
1744350 c.757C>A p.P253T Chr.6: 134172306-134172306 on Build GRCh38
1073542 c.978T>A p.D326E Chr.6: 134172286-134172286 on Build GRCh38
450533 c.980C>A p.S327* Chr.6: 134172284-134172284 on Build GRCh38
2152533 c.693C>T p.F231F Chr.6: 134172256-134172256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55811643 Chr.6: 134172329-134172333 on Build GRCh38
rs894495021 Chr.6: 134172332-134172332 on Build GRCh38
rs747061027 Chr.6: 134172321-134172321 on Build GRCh38
rs760092718 Chr.6: 134172324-134172324 on Build GRCh38
rs375679330 Chr.6: 134172365-134172365 on Build GRCh38
rs777389292 Chr.6: 134172374-134172374 on Build GRCh38
rs758675767 Chr.6: 134172317-134172318 on Build GRCh38
rs752706271 Chr.6: 134172346-134172346 on Build GRCh38
rs755939511 Chr.6: 134172326-134172326 on Build GRCh38
rs761026399 Chr.6: 134172350-134172350 on Build GRCh38
rs150642168 Chr.6: 134172312-134172312 on Build GRCh38
rs781574579 Chr.6: 134172337-134172339 on Build GRCh38
rs772039358 Chr.6: 134172344-134172345 on Build GRCh38
rs778063439 Chr.6: 134172320-134172321 on Build GRCh38
rs748017172 Chr.6: 134172292-134172292 on Build GRCh38
rs141890757 Chr.6: 134172273-134172273 on Build GRCh38
rs773691115 Chr.6: 134172286-134172286 on Build GRCh38
rs746720665 Chr.6: 134172285-134172285 on Build GRCh38
rs184691170 Chr.6: 134172361-134172361 on Build GRCh38
rs763411927 Chr.6: 134172291-134172291 on Build GRCh38

View Full Product Details