Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56463537-56463703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
UniGene
Hs.604915
Gene Chromosome Location
Chr.6: 56457987-56954671 on Build GRCh38
Transcripts
NM_001144769.2 NM_001144770.1 NM_015548.4 NM_183380.3 XM_005249310.3 XM_005249315.3 XM_005249316.2 XM_005249318.2 XM_005249319.2 XM_005249320.4 XM_005249322.4 XM_005249323.3 XM_005249324.2 XM_011514824.2 XM_011514825.2 XM_011514826.2 XM_017011205.1 XM_017011206.1 XM_017011207.1 XM_017011208.1 XM_017011209.1 XM_017011210.1 XM_017011211.1 XM_017011212.1 XM_017011213.1 XM_017011214.1 XM_017011215.1 XM_017011216.1 XM_017011217.1 XM_017011218.1 XM_017011219.1 XM_017011220.1 XM_017011221.1 XM_017011222.1 XM_017011223.1 XM_017011224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5450276 c.14994G>T p.Q4998H Chr.6: 56463589-56463589 on Build GRCh38
3628951 c.14999C>T p.P5000L Chr.6: 56463584-56463584 on Build GRCh38
3874919 c.22145G>T p.R7382I Chr.6: 56463671-56463671 on Build GRCh38
3628952 c.22232C>T p.P7411L Chr.6: 56463584-56463584 on Build GRCh38
3874917 c.22150C>T p.R7384W Chr.6: 56463666-56463666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771999169 Chr.6: 56463648-56463648 on Build GRCh38
rs750916776 Chr.6: 56463602-56463602 on Build GRCh38
rs200781365 Chr.6: 56463628-56463628 on Build GRCh38
rs748716722 Chr.6: 56463627-56463627 on Build GRCh38
rs763551194 Chr.6: 56463682-56463682 on Build GRCh38
rs201293893 Chr.6: 56463579-56463579 on Build GRCh38
rs377586570 Chr.6: 56463664-56463664 on Build GRCh38
rs375303601 Chr.6: 56463562-56463562 on Build GRCh38
rs202108973 Chr.6: 56463586-56463586 on Build GRCh38
rs776951210 Chr.6: 56463666-56463666 on Build GRCh38
rs777534699 Chr.6: 56463624-56463624 on Build GRCh38
rs760819112 Chr.6: 56463569-56463569 on Build GRCh38
rs374094129 Chr.6: 56463592-56463592 on Build GRCh38
rs370820734 Chr.6: 56463667-56463667 on Build GRCh38
rs557488147 Chr.6: 56463570-56463570 on Build GRCh38
rs554371778 Chr.6: 56463601-56463601 on Build GRCh38
rs775275480 Chr.6: 56463568-56463568 on Build GRCh38
rs948596983 Chr.6: 56463681-56463681 on Build GRCh38
rs768734906 Chr.6: 56463665-56463665 on Build GRCh38
rs745750653 Chr.6: 56463647-56463647 on Build GRCh38
rs201429821 Chr.6: 56463608-56463608 on Build GRCh38
rs201785384 Chr.6: 56463653-56463653 on Build GRCh38
rs750542445 Chr.6: 56463680-56463680 on Build GRCh38
rs765310351 Chr.6: 56463674-56463674 on Build GRCh38
rs901364111 Chr.6: 56463632-56463632 on Build GRCh38
rs775260497 Chr.6: 56463655-56463655 on Build GRCh38
rs200298542 Chr.6: 56463618-56463618 on Build GRCh38
rs201952309 Chr.6: 56463585-56463585 on Build GRCh38
rs778309897 Chr.6: 56463629-56463629 on Build GRCh38
rs567935553 Chr.6: 56463607-56463607 on Build GRCh38

View Full Product Details