Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 61732660-61732815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL3
Entrez Gene ID
23284
Gene Name
adhesion G protein-coupled receptor L3
Gene Aliases
CIRL3, CL3, LEC3, LPHN3
UniGene
Hs.28391
Gene Chromosome Location
Chr.4: 61201256-62078335 on Build GRCh38
Transcripts
NM_001322246.1 NM_001322402.1 NM_015236.5 XM_011531788.2 XM_011531791.2 XM_017007929.1 XM_017007930.1 XM_017007931.1 XM_017007932.1 XM_017007933.1 XM_017007934.1 XM_017007935.1 XM_017007936.1 XM_017007937.1 XM_017007938.1 XM_017007939.1 XM_017007940.1 XM_017007941.1 XM_017007942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4610616 c.400C>A p.L134I Chr.4: 61732759-61732759 on Build GRCh38
4610617 c.604C>A p.L202I Chr.4: 61732759-61732759 on Build GRCh38
402450 c.410G>T p.G137V Chr.4: 61732769-61732769 on Build GRCh38
6041354 c.657C>T p.S219S Chr.4: 61732812-61732812 on Build GRCh38
6041353 c.453C>T p.S151S Chr.4: 61732812-61732812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893106568 Chr.4: 61732693-61732693 on Build GRCh38
rs750359901 Chr.4: 61732783-61732783 on Build GRCh38
rs762354956 Chr.4: 61732720-61732723 on Build GRCh38
rs576366112 Chr.4: 61732726-61732726 on Build GRCh38
rs1013435696 Chr.4: 61732711-61732711 on Build GRCh38
rs753087657 Chr.4: 61732699-61732699 on Build GRCh38
rs765037132 Chr.4: 61732698-61732698 on Build GRCh38
rs776711960 Chr.4: 61732747-61732748 on Build GRCh38

View Full Product Details