Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43281974-43282117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGM7
Entrez Gene ID
116179
Gene Name
transglutaminase 7
Gene Aliases
TGMZ
UniGene
Hs.164661
Gene Chromosome Location
Chr.15: 43276277-43302255 on Build GRCh38
Transcripts
NM_052955.2 XM_017021903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2188070 c.1170C>A p.A390A Chr.15: 43282025-43282025 on Build GRCh38
3816118 c.1199T>G p.V400G Chr.15: 43281996-43281996 on Build GRCh38
5383350 c.1180C>T p.P394S Chr.15: 43282015-43282015 on Build GRCh38
2188073 c.1130C>T p.A377V Chr.15: 43282065-43282065 on Build GRCh38
2188072 c.1155G>A p.G385G Chr.15: 43282040-43282040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761067229 Chr.15: 43282047-43282047 on Build GRCh38
rs772892631 Chr.15: 43281998-43281998 on Build GRCh38
rs776023747 Chr.15: 43282022-43282022 on Build GRCh38
rs764432955 Chr.15: 43282065-43282065 on Build GRCh38
rs770413654 Chr.15: 43282006-43282006 on Build GRCh38
rs530951588 Chr.15: 43282017-43282017 on Build GRCh38
rs1017676423 Chr.15: 43282039-43282039 on Build GRCh38
rs754076313 Chr.15: 43282077-43282077 on Build GRCh38
rs267604216 Chr.15: 43282025-43282025 on Build GRCh38
rs767875545 Chr.15: 43282062-43282066 on Build GRCh38
rs773851368 Chr.15: 43282014-43282014 on Build GRCh38
rs957049082 Chr.15: 43282060-43282060 on Build GRCh38
rs753018352 Chr.15: 43282067-43282067 on Build GRCh38
rs768030689 Chr.15: 43282019-43282019 on Build GRCh38
rs762583560 Chr.15: 43282000-43282000 on Build GRCh38
rs1009007198 Chr.15: 43282034-43282034 on Build GRCh38
rs769530051 Chr.15: 43281994-43281994 on Build GRCh38
rs757435223 Chr.15: 43282087-43282087 on Build GRCh38
rs111755161 Chr.15: 43282018-43282018 on Build GRCh38

View Full Product Details