Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 89127110-89127281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA3
Entrez Gene ID
2042
Gene Name
EPH receptor A3
Gene Aliases
EK4, ETK, ETK1, HEK, HEK4, TYRO4
UniGene
Hs.123642
Gene Chromosome Location
Chr.3: 89107524-89482134 on Build GRCh38
Transcripts
NM_005233.5 NM_182644.2 XM_005264715.2 XM_005264716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4120473 c.98T>G p.L33R Chr.3: 89127218-89127218 on Build GRCh38
4120472 c.98T>G p.L33R Chr.3: 89127218-89127218 on Build GRCh38
5059978 c.101A>G p.D34G Chr.3: 89127221-89127221 on Build GRCh38
731481 c.137C>G p.S46C Chr.3: 89127257-89127257 on Build GRCh38
1425619 c.109A>G p.T37A Chr.3: 89127229-89127229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755864850 Chr.3: 89127168-89127168 on Build GRCh38
rs756018331 Chr.3: 89127245-89127245 on Build GRCh38
rs372126385 Chr.3: 89127206-89127206 on Build GRCh38
rs372273179 Chr.3: 89127248-89127248 on Build GRCh38
rs560245157 Chr.3: 89127195-89127195 on Build GRCh38
rs760801471 Chr.3: 89127235-89127235 on Build GRCh38
rs367884951 Chr.3: 89127197-89127197 on Build GRCh38
rs766626719 Chr.3: 89127237-89127237 on Build GRCh38
rs374211029 Chr.3: 89127241-89127241 on Build GRCh38
rs950929609 Chr.3: 89127261-89127261 on Build GRCh38
rs757449877 Chr.3: 89127182-89127182 on Build GRCh38
rs748693901 Chr.3: 89127202-89127202 on Build GRCh38
rs368633855 Chr.3: 89127247-89127247 on Build GRCh38
rs773685257 Chr.3: 89127216-89127216 on Build GRCh38
rs139262347 Chr.3: 89127255-89127255 on Build GRCh38
rs759710031 Chr.3: 89127244-89127244 on Build GRCh38
rs368848126 Chr.3: 89127183-89127183 on Build GRCh38
rs138358922 Chr.3: 89127184-89127184 on Build GRCh38
rs778918699 Chr.3: 89127155-89127177 on Build GRCh38
rs374879413 Chr.3: 89127196-89127196 on Build GRCh38

View Full Product Details