Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 140233081-140233246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP1B
Entrez Gene ID
53353
Gene Name
LDL receptor related protein 1B
Gene Aliases
LRP-1B, LRP-DIT, LRPDIT
UniGene
Hs.656461
Gene Chromosome Location
Chr.2: 140231423-142132463 on Build GRCh38
Transcripts
NM_018557.2 XM_017004341.1 XM_017004342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1723593 c.13742A>G p.E4581G Chr.2: 140233244-140233244 on Build GRCh38
4698820 c.13798T>C p.*4600Q Chr.2: 140233188-140233188 on Build GRCh38
3567095 c.13764G>A p.K4588K Chr.2: 140233222-140233222 on Build GRCh38
7208014 c.13766A>T p.K4589I Chr.2: 140233220-140233220 on Build GRCh38
475954 c.13746G>A p.R4582R Chr.2: 140233240-140233240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147658745 Chr.2: 140233156-140233156 on Build GRCh38
rs748901705 Chr.2: 140233212-140233212 on Build GRCh38
rs754176419 Chr.2: 140233211-140233212 on Build GRCh38
rs754051799 Chr.2: 140233146-140233146 on Build GRCh38
rs894108871 Chr.2: 140233208-140233208 on Build GRCh38
rs758134771 Chr.2: 140233150-140233150 on Build GRCh38
rs755036642 Chr.2: 140233169-140233169 on Build GRCh38
rs940516254 Chr.2: 140233117-140233117 on Build GRCh38
rs557818790 Chr.2: 140233179-140233179 on Build GRCh38
rs777318122 Chr.2: 140233206-140233206 on Build GRCh38
rs375100156 Chr.2: 140233162-140233162 on Build GRCh38
rs142353749 Chr.2: 140233187-140233187 on Build GRCh38
rs972248186 Chr.2: 140233141-140233141 on Build GRCh38
rs755032750 Chr.2: 140233217-140233217 on Build GRCh38

View Full Product Details