Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47372947-47373091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMPK1
Entrez Gene ID
51727
Gene Name
cytidine/uridine monophosphate kinase 1
Gene Aliases
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK
UniGene
Hs.731647
Gene Chromosome Location
Chr.1: 47333797-47378839 on Build GRCh38
Transcripts
NM_001136140.1 NM_016308.2 NR_046394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2191781 c.326A>T p.D109V Chr.1: 47372962-47372962 on Build GRCh38
910429 c.363C>A p.F121L Chr.1: 47372999-47372999 on Build GRCh38
2191783 c.354G>T p.K118N Chr.1: 47372990-47372990 on Build GRCh38
146505 c.344A>G p.N115S Chr.1: 47372980-47372980 on Build GRCh38
4428419 c.331A>G p.T111A Chr.1: 47372967-47372967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376110727 Chr.1: 47372979-47372979 on Build GRCh38
rs762579970 Chr.1: 47373049-47373049 on Build GRCh38
rs763787616 Chr.1: 47373052-47373052 on Build GRCh38
rs775071933 Chr.1: 47372986-47372986 on Build GRCh38
rs892812530 Chr.1: 47373041-47373041 on Build GRCh38
rs960886197 Chr.1: 47372994-47372994 on Build GRCh38
rs977859310 Chr.1: 47372973-47372973 on Build GRCh38
rs72553947 Chr.1: 47372980-47372980 on Build GRCh38
rs1054097379 Chr.1: 47373022-47373022 on Build GRCh38
rs751273816 Chr.1: 47373057-47373057 on Build GRCh38

View Full Product Details