Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55479465-55479630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP2
Entrez Gene ID
4313
Gene Name
matrix metallopeptidase 2
Gene Aliases
CLG4, CLG4A, MMP-2, MMP-II, MONA, TBE-1
UniGene
Hs.513617
Gene Chromosome Location
Chr.16: 55478830-55506691 on Build GRCh38
Transcripts
NM_001302508.1 NM_004530.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956163 c.75C>T p.S25S Chr.16: 55479554-55479554 on Build GRCh38
4702695 c.8C>T p.A3V Chr.16: 55479487-55479487 on Build GRCh38
5454457 c.27G>T p.A9A Chr.16: 55479506-55479506 on Build GRCh38
191656 c.96G>A p.S32S Chr.16: 55479575-55479575 on Build GRCh38
3667868 c.58C>T p.L20L Chr.16: 55479537-55479537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770535923 Chr.16: 55479549-55479549 on Build GRCh38
rs746268212 Chr.16: 55479499-55479499 on Build GRCh38
rs781428467 Chr.16: 55479491-55479491 on Build GRCh38
rs781581204 Chr.16: 55479502-55479502 on Build GRCh38
rs998400659 Chr.16: 55479567-55479567 on Build GRCh38
rs904959190 Chr.16: 55479497-55479497 on Build GRCh38
rs769217964 Chr.16: 55479532-55479532 on Build GRCh38
rs764055331 Chr.16: 55479584-55479584 on Build GRCh38
rs868623056 Chr.16: 55479488-55479488 on Build GRCh38
rs776312366 Chr.16: 55479553-55479553 on Build GRCh38
rs146220690 Chr.16: 55479575-55479575 on Build GRCh38
rs751615898 Chr.16: 55479586-55479586 on Build GRCh38
rs747046465 Chr.16: 55479484-55479484 on Build GRCh38
rs780763772 Chr.16: 55479516-55479516 on Build GRCh38
rs757454010 Chr.16: 55479489-55479489 on Build GRCh38
rs760180233 Chr.16: 55479548-55479548 on Build GRCh38
rs41503347 Chr.16: 55479530-55479530 on Build GRCh38
rs770252807 Chr.16: 55479508-55479508 on Build GRCh38
rs765007075 Chr.16: 55479561-55479561 on Build GRCh38
rs139082366 Chr.16: 55479596-55479596 on Build GRCh38
rs1016960822 Chr.16: 55479603-55479603 on Build GRCh38
rs575398965 Chr.16: 55479572-55479572 on Build GRCh38
rs243863 Chr.16: 55479558-55479558 on Build GRCh38
rs772627742 Chr.16: 55479542-55479542 on Build GRCh38

View Full Product Details