Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138515134-138515308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM24
Entrez Gene ID
8805
Gene Name
tripartite motif containing 24
Gene Aliases
PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1
UniGene
Hs.490287
Gene Chromosome Location
Chr.7: 138460334-138585588 on Build GRCh38
Transcripts
NM_003852.3 NM_015905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5666744 c.517A>C p.N173H Chr.7: 138515245-138515245 on Build GRCh38
7167494 c.433T>A p.C145S Chr.7: 138515287-138515287 on Build GRCh38
4735983 c.508G>A p.A170T Chr.7: 138515236-138515236 on Build GRCh38
5666745 c.517A>C p.N173H Chr.7: 138515245-138515245 on Build GRCh38
5666746 c.391A>C p.N131H Chr.7: 138515245-138515245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149374418 Chr.7: 138515285-138515285 on Build GRCh38
rs759796188 Chr.7: 138515245-138515245 on Build GRCh38
rs146342480 Chr.7: 138515286-138515286 on Build GRCh38
rs1044884465 Chr.7: 138515220-138515220 on Build GRCh38
rs770482864 Chr.7: 138515235-138515235 on Build GRCh38
rs768691550 Chr.7: 138515187-138515187 on Build GRCh38
rs757539755 Chr.7: 138515167-138515167 on Build GRCh38
rs753104674 Chr.7: 138515262-138515262 on Build GRCh38
rs986393126 Chr.7: 138515197-138515197 on Build GRCh38
rs573488388 Chr.7: 138515172-138515172 on Build GRCh38
rs762243595 Chr.7: 138515163-138515163 on Build GRCh38
rs749006983 Chr.7: 138515182-138515182 on Build GRCh38
rs765793542 Chr.7: 138515246-138515246 on Build GRCh38
rs772860612 Chr.7: 138515196-138515196 on Build GRCh38
rs201699567 Chr.7: 138515226-138515226 on Build GRCh38
rs542483128 Chr.7: 138515190-138515190 on Build GRCh38
rs367622117 Chr.7: 138515191-138515191 on Build GRCh38
rs771506519 Chr.7: 138515195-138515195 on Build GRCh38
rs777090182 Chr.7: 138515236-138515236 on Build GRCh38
rs892912620 Chr.7: 138515157-138515157 on Build GRCh38
rs760373763 Chr.7: 138515214-138515214 on Build GRCh38

View Full Product Details