Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35930155-35930380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RDM1
Entrez Gene ID
201299
Gene Name
RAD52 motif containing 1
Gene Aliases
RAD52B
UniGene
Hs.194411
Gene Chromosome Location
Chr.17: 35918080-35931821 on Build GRCh38
Transcripts
NM_001034836.1 NM_001163120.1 NM_001163121.1 NM_001163122.1 NM_001163124.1 NM_001163125.1 NM_001163130.1 NM_145654.3 NR_027996.1 NR_027997.1 NR_027998.1 NR_027999.1 NR_028000.1 XM_011524509.2 XM_011524510.2

SNP Detail

SNP ID Chromosome Location
rs999659398 Chr.17: 35930339-35930339 on Build GRCh38
rs9907295 Chr.17: 35930309-35930309 on Build GRCh38
rs775396568 Chr.17: 35930292-35930292 on Build GRCh38
rs971088303 Chr.17: 35930325-35930325 on Build GRCh38
rs200333515 Chr.17: 35930260-35930260 on Build GRCh38
rs768701786 Chr.17: 35930297-35930297 on Build GRCh38
rs1006399693 Chr.17: 35930211-35930211 on Build GRCh38
rs1039656984 Chr.17: 35930216-35930216 on Build GRCh38
rs771121905 Chr.17: 35930287-35930287 on Build GRCh38
rs375786999 Chr.17: 35930230-35930231 on Build GRCh38
rs772161923 Chr.17: 35930340-35930340 on Build GRCh38
rs755785027 Chr.17: 35930223-35930223 on Build GRCh38
rs766587085 Chr.17: 35930271-35930271 on Build GRCh38
rs149206374 Chr.17: 35930202-35930202 on Build GRCh38
rs749590532 Chr.17: 35930256-35930256 on Build GRCh38
rs756818417 Chr.17: 35930195-35930195 on Build GRCh38
rs772152117 Chr.17: 35930282-35930282 on Build GRCh38
rs1000850853 Chr.17: 35930255-35930255 on Build GRCh38
rs958076779 Chr.17: 35930342-35930342 on Build GRCh38
rs577071383 Chr.17: 35930264-35930264 on Build GRCh38
rs766177028 Chr.17: 35930248-35930249 on Build GRCh38
rs750107163 Chr.17: 35930205-35930205 on Build GRCh38
rs767169918 Chr.17: 35930329-35930329 on Build GRCh38
rs369831618 Chr.17: 35930293-35930293 on Build GRCh38
rs183791835 Chr.17: 35930307-35930307 on Build GRCh38
rs748284868 Chr.17: 35930270-35930270 on Build GRCh38
rs758171819 Chr.17: 35930188-35930188 on Build GRCh38
rs773207129 Chr.17: 35930283-35930283 on Build GRCh38
rs767081884 Chr.17: 35930199-35930199 on Build GRCh38
rs779283337 Chr.17: 35930263-35930263 on Build GRCh38
rs769890154 Chr.17: 35930229-35930229 on Build GRCh38
rs375253320 Chr.17: 35930286-35930286 on Build GRCh38
rs1042242676 Chr.17: 35930189-35930189 on Build GRCh38

View Full Product Details