Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48845483-48845708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHA4
Entrez Gene ID
57664
Gene Name
pleckstrin homology domain containing A4
Gene Aliases
PEPP1
UniGene
Hs.9469
Gene Chromosome Location
Chr.19: 48837092-48868627 on Build GRCh38
Transcripts
NM_001161354.1 NM_020904.2 XM_005259107.2 XM_006723301.2 XM_011527157.1 XM_011527158.2 XM_011527159.1 XM_011527160.1 XM_011527162.2 XM_017027037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3536676 c.1630C>T p.P544S Chr.19: 48845553-48845553 on Build GRCh38
4601560 c.1624C>T p.R542W Chr.19: 48845559-48845559 on Build GRCh38
3536677 c.1597C>T p.P533S Chr.19: 48845586-48845586 on Build GRCh38
999092 c.1666G>A p.G556S Chr.19: 48845517-48845517 on Build GRCh38
4469339 c.1595C>T p.S532F Chr.19: 48845588-48845588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960241070 Chr.19: 48845588-48845588 on Build GRCh38
rs758048584 Chr.19: 48845533-48845533 on Build GRCh38
rs1014723986 Chr.19: 48845603-48845603 on Build GRCh38
rs774254686 Chr.19: 48845672-48845672 on Build GRCh38
rs770104741 Chr.19: 48845653-48845653 on Build GRCh38
rs199687390 Chr.19: 48845514-48845514 on Build GRCh38
rs752535120 Chr.19: 48845595-48845595 on Build GRCh38
rs957914203 Chr.19: 48845651-48845651 on Build GRCh38
rs61755449 Chr.19: 48845585-48845585 on Build GRCh38
rs541379003 Chr.19: 48845659-48845659 on Build GRCh38
rs745920733 Chr.19: 48845559-48845559 on Build GRCh38
rs925074523 Chr.19: 48845654-48845656 on Build GRCh38
rs201390061 Chr.19: 48845621-48845621 on Build GRCh38
rs142897659 Chr.19: 48845572-48845572 on Build GRCh38
rs757489084 Chr.19: 48845555-48845555 on Build GRCh38
rs764590410 Chr.19: 48845512-48845512 on Build GRCh38
rs762138777 Chr.19: 48845573-48845573 on Build GRCh38
rs748513152 Chr.19: 48845652-48845652 on Build GRCh38
rs772191142 Chr.19: 48845561-48845561 on Build GRCh38
rs769190377 Chr.19: 48845637-48845637 on Build GRCh38
rs145653699 Chr.19: 48845544-48845544 on Build GRCh38
rs779178027 Chr.19: 48845607-48845607 on Build GRCh38
rs773986812 Chr.19: 48845663-48845663 on Build GRCh38
rs555571700 Chr.19: 48845620-48845620 on Build GRCh38
rs986719636 Chr.19: 48845591-48845591 on Build GRCh38
rs766385316 Chr.19: 48845537-48845537 on Build GRCh38
rs768938383 Chr.19: 48845566-48845566 on Build GRCh38
rs866459224 Chr.19: 48845527-48845527 on Build GRCh38
rs1020394140 Chr.19: 48845610-48845610 on Build GRCh38
rs375680300 Chr.19: 48845562-48845562 on Build GRCh38
rs200502456 Chr.19: 48845532-48845532 on Build GRCh38
rs755905495 Chr.19: 48845605-48845605 on Build GRCh38
rs879442631 Chr.19: 48845668-48845668 on Build GRCh38
rs144775663 Chr.19: 48845574-48845574 on Build GRCh38
rs779074447 Chr.19: 48845558-48845558 on Build GRCh38
rs28455995 Chr.19: 48845638-48845638 on Build GRCh38
rs539628897 Chr.19: 48845545-48845545 on Build GRCh38
rs770508236 Chr.19: 48845662-48845662 on Build GRCh38
rs925192976 Chr.19: 48845648-48845648 on Build GRCh38
rs372118677 Chr.19: 48845655-48845655 on Build GRCh38
rs140457819 Chr.19: 48845538-48845538 on Build GRCh38
rs556416279 Chr.19: 48845611-48845611 on Build GRCh38
rs767414198 Chr.19: 48845590-48845590 on Build GRCh38
rs772777371 Chr.19: 48845575-48845575 on Build GRCh38
rs566414042 Chr.19: 48845568-48845568 on Build GRCh38
rs766008964 Chr.19: 48845581-48845581 on Build GRCh38
rs747542732 Chr.19: 48845565-48845565 on Build GRCh38
rs762529557 Chr.19: 48845580-48845580 on Build GRCh38
rs1004580664 Chr.19: 48845567-48845567 on Build GRCh38

View Full Product Details